Skip to Content
Merck
All Photos(3)

Key Documents

215406

Sigma-Aldrich

Ammonium iron(II) sulfate hexahydrate

ACS reagent, 99%

Synonym(s):

Ammonium ferrous sulfate hexahydrate, Ammonium iron(II) sulfate, Mohr’s salt

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(NH4)2Fe(SO4)2 · 6H2O
CAS Number:
Molecular Weight:
392.14
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NB.24
Assay:
98.5-101.5%
99%
grade:
ACS reagent
form:
powder or crystals
Pricing and availability is not currently available.

grade

ACS reagent

Quality Level

Assay

98.5-101.5%
99%

form

powder or crystals

reaction suitability

reagent type: catalyst
core: iron

impurities

≤0.01% insolubles
≤0.05% Not NH4OH pptd.

pH

3-5 (20 °C, 50 g/L)

mp

100 °C (dec.) (lit.)

anion traces

phosphate (PO43-): ≤0.003%

cation traces

Ca: 0.005%
Cu: ≤0.003%
Fe3+: ≤0.01%
K: 0.002%
Mg: 0.002%
Mn: ≤0.01%
Na: 0.02%
Zn: ≤0.003%

SMILES string

N.N.O.O.O.O.O.O.[Fe++].OS([O-])(=O)=O.OS([O-])(=O)=O

InChI

1S/Fe.2H3N.2H2O4S.6H2O/c;;;2*1-5(2,3)4;;;;;;/h;2*1H3;2*(H2,1,2,3,4);6*1H2/q+2;;;;;;;;;;/p-2

InChI key

MQLVWQSVRZVNIP-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Application

Ammonium iron(II) sulfate hexahydrate can be used as a green catalyst in the:
  • Newman-Kwart rearrangement to synthesize functionalized thiophenols.
  • Oxidation of aromatic, aliphatic, and conjugated aldehydes to the corresponding carboxylic acids.
  • Synthesis of biscoumarins via condensation reaction.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Konstantinos Grintzalis et al.
Free radical biology & medicine, 59, 27-35 (2012-10-09)
A simple and sensitive method is presented for the simultaneous quantification (spectrophotometric and spectrofluorimetric) of the main lipid and protein peroxidation products after their initial fractionation: free malondialdehyde (FrMDA), protein-bound malondialdehyde (PrMDA), total hydroperoxides (LOOH), and protein hydroperoxides (PrOOH). FrMDA
Metallization of biologically inspired silica nanotubes.
Jo W, et al.
Materials Science and Engineering, C, 32(8), 2426-2430 (2012)
Mixed ligand complexes of iron (II) and (III) with cyanide and aromatic di-imines.
Schilt AA.
Journal of the American Chemical Society, 82(12), 3000-3005 (1960)
The crystal structure of Tutton's salts. VI. Vanadium (II), iron (II) and cobalt (II) ammonium sulfate hexahydrates.
Montgomery H, et al.
Acta Crystallographica, 22(6), 775-780 (1967)
Apoorvi Tyagi et al.
International journal of molecular sciences, 21(23) (2020-12-03)
Phenylketonuria (PKU) is an autosomal recessive metabolic disorder caused by the dysfunction of the enzyme phenylalanine hydroxylase (PAH). Alterations in the level of PAH leads to the toxic accumulation of phenylalanine in the blood and brain. Protein degradation mediated by

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service