Skip to Content
Merck
All Photos(1)

Key Documents

102466

Sigma-Aldrich

Ethylene glycol

ReagentPlus®, ≥99%

Synonym(s):

1,2-Ethanediol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HOCH2CH2OH
CAS Number:
Molecular Weight:
62.07
Beilstein:
505945
EC Number:
MDL number:
UNSPSC Code:
12352137
eCl@ss:
39020501
PubChem Substance ID:
NACRES:
NA.21
grade:
reagent
bp:
195-198 °C
vapor pressure:
0.08 mmHg ( 20 °C)
Pricing and availability is not currently available.

grade

reagent

Quality Level

vapor density

2.1 (vs air)

vapor pressure

0.08 mmHg ( 20 °C)

product line

ReagentPlus®

Assay

≥99%

form

liquid

autoignition temp.

752 °F

expl. lim.

15.3 %

dilution

(for general lab use)

refractive index

n20/D 1.431 (lit.)

bp

195-198 °C

mp

−13 °C (lit.)

solubility

water: miscible

density

1.113 g/mL at 25 °C (lit.)

storage temp.

room temp

SMILES string

OCCO

InChI

1S/C2H6O2/c3-1-2-4/h3-4H,1-2H2

InChI key

LYCAIKOWRPUZTN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Ethylene glycol (EG) is a polyol having a wide range of industrial applications. Its preparation from cellulose has been reported.[1] It can also be produced from fossil fuels (petroleum, natural gas and coal) and biomass-derived resources.[2]

Application

Ethylene glycol can be used as a reagent in cyclocondensation reactions with aldehydes and ketones to form 1,3-dioxolanes.[3][4]
It has been used:
  • As a carrier fluid during the fabrication of shear thickening fluid (composed of nanosized silica particles).[5]
  • To compose previtrification and vitrification solutions for the cryopreservation of mouse embryos.[6]

Other Notes

The article number 102466-4X2.5L will be discontinued. Please order the single bottle 102466-2.5L which is physically identical with the same exact specifications.

Legal Information

ReagentPlus is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Health hazardExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - STOT RE 2 Oral

Target Organs

Kidney

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

239.0 °F - open cup

Flash Point(C)

115 °C - open cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Siloxanes as temporary tethers in [2+2] photocycloadditions
Crimmins MT and Guise LE
Tetrahedron Letters, 35(11), 1657-1660 (1994)
Study on magnetorheological shear thickening fluid.
Zhang X, et al.
Smart Materials and Structures, 17(1), 015051-015051 (2008)
Sulfonated charcoal, a mild and efficient reagent for the preparation of cyclic acetals, dithioacetals and benzodioxepines
Patney HK
Tetrahedron Letters, 32(3), 413-416 (1991)
Mouse embryo cryopreservation utilizing a novel high-capacity vitrification spatula.
Tsang WH and Chow KL.
Biotechniques, 46, 550-552 (2009)
Ethylene glycol: properties, synthesis, and applications.
Yue H, et al.
Chemical Society Reviews, 41(11), 4218-4244 (2012)

Related Content

High-throughput reagents for drug target screening and immunogenicity testing.

High-throughput reagents for drug target screening and immunogenicity testing.

High-throughput reagents for drug target screening and immunogenicity testing.

High-throughput reagents for drug target screening and immunogenicity testing.

Questions

1–3 of 3 Questions  
  1. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

  2. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

  3. Is this product sterile and usable for cell culture?

    1 answer
    1. This product has not been tested for sterility or for use in cell culture. However, based on several published articles, it has been previously used in cell culture applications. The end-user will need to determine suitability.

      Please use the link below to review the peer-reviewed articles associated with this product:
      https://www.sigmaaldrich.com/search/102466?focus=papers&page=1&perpage=30&sort=relevance&term=102466&type=citation_search

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service