Skip to Content
Merck
All Photos(1)

Documents

Y0000796

Fluorescein

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

Acid Yellow 73

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H12O5
CAS Number:
Molecular Weight:
332.31
Colour Index Number:
45350
Beilstein:
94324
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

Agency

EP Reference Standard

API family

fluorescein

manufacturer/tradename

EDQM

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

Oc1ccc2c(Oc3cc(O)ccc3C24OC(=O)c5ccccc45)c1

InChI

1S/C20H12O5/c21-11-5-7-15-17(9-11)24-18-10-12(22)6-8-16(18)20(15)14-4-2-1-3-13(14)19(23)25-20/h1-10,21-22H

InChI key

GNBHRKFJIUUOQI-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Fluorescein EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Wout O Rohof et al.
Clinical gastroenterology and hepatology : the official clinical practice journal of the American Gastroenterological Association, 12(10), 1647-1655 (2013-11-05)
Approximately 30% of patients with gastroesophageal reflux disease have symptoms resistant to treatment with proton pump inhibitors (PPIs). Several mechanisms such as esophageal hypersensitivity, increased mucosal permeability, and possibly the position of the gastric acid pocket might underlie a partial
Thaned Kangsamaksin et al.
Cancer discovery, 5(2), 182-197 (2014-11-13)
A proangiogenic role for Jagged (JAG)-dependent activation of NOTCH signaling in the endothelium has yet to be described. Using proteins that encoded different NOTCH1 EGF-like repeats, we identified unique regions of Delta-like ligand (DLL)-class and JAG-class ligand-receptor interactions, and developed
Tomohisa Akamatsu et al.
The American journal of pathology, 184(6), 1843-1852 (2014-04-16)
Neonatal hypoxic-ischemic encephalopathy (HIE) remains a serious burden in neonatal care. Hypothermia provides a good outcome in some babies with HIE. Here, we investigated the biological mechanisms of its neuroprotective effect and sought for a new therapeutic target. We made
Osaama H Khan et al.
Neurosurgical focus, 37(4), E7-E7 (2014-12-04)
This paper describes a consecutive series of skull base meningiomas resected using an endoscopic endonasal approach through various corridors at a single institution over 7 years. The impact of case selection and experience, the presence of a cortical cuff between
Aditya Venkatesh et al.
The Journal of clinical investigation, 125(4), 1446-1458 (2015-03-24)
Retinitis pigmentosa (RP) is an inherited photoreceptor degenerative disorder that results in blindness. The disease is often caused by mutations in genes that are specific to rod photoreceptors; however, blindness results from the secondary loss of cones by a still

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service