Skip to Content
Merck
All Photos(1)

Key Documents

Y0000766

Phthalic acid

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

1,2-Benzenedicarboxylic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H4-1,2-(CO2H)2
CAS Number:
Molecular Weight:
166.13
Beilstein:
608199
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24
Pricing and availability is not currently available.

grade

pharmaceutical primary standard

API family

fluorescein

manufacturer/tradename

EDQM

mp

210-211 °C (dec.) (lit.)

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

OC(C1=C(C(O)=O)C=CC=C1)=O

InChI

1S/C8H6O4/c9-7(10)5-3-1-2-4-6(5)8(11)12/h1-4H,(H,9,10)(H,11,12)

InChI key

XNGIFLGASWRNHJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Phthalic acid EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Dam. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

334.4 °F

Flash Point(C)

168 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mahmoud M Abdel daiem et al.
Journal of environmental management, 109, 164-178 (2012-07-17)
This article describes the most recent methods developed to remove phthalic acid esters (PAEs) from water, wastewater, sludge, and soil. In general, PAEs are considered to be endocrine disrupting chemicals (EDCs), whose effects may not appear until long after exposure.
Ying Zhang et al.
The Science of the total environment, 506-507, 118-125 (2014-12-03)
The current study investigates the existence of 15 phthalate esters (PAEs) in surface soils (27 samples) collected from 9 different facility agriculture sites in the black soil region of northeast China, during the process of agricultural production (comprising only three
En-Cui Yang et al.
Dalton transactions (Cambridge, England : 2003), 44(7), 3190-3199 (2015-01-13)
Four new 4-amino-1,2,4-triazole (atr)-based coordination polymers, {[Cu2(atr)(H2O)(μ-OH)2(pa)]·H2O}n (), {[Cu3(atr)2(H2O)2(μ-OH)2(npa)2]·2H2O}n (), {[Cu3(atr)5(dca)(μ-OH)(ClO4)2](ClO4)2}n () and {[Cu3(atr)2(H2O)(μ3-OH)(μ-OH)2(spa)]·1.5H2O}n () (pa(2-) = phthalate, npa(2-) = 3-nitrophthalate, dca(-) = dicyanamide and spa(3-) = 4-sulfophthalate), were successfully obtained by varying the carboxylate- and cyanide-modified magnetic bridges. Structural
Xiao-Yan Zhang et al.
Applied and environmental microbiology, 80(22), 6870-6878 (2014-08-26)
EstS1, a newly identified thermostable esterase from Sulfobacillus acidophilus DSM10332, was heterologously expressed in Escherichia coli and shown to enzymatically degrade phthalate esters (PAEs) to their corresponding monoalkyl PAEs. The optimal pH and temperature of the esterase were found to
Germaine M Buck Louis et al.
Fertility and sterility, 101(5), 1359-1366 (2014-02-19)
To assess the relationship between environmental chemicals and couple fecundity or time to pregnancy (TTP). Prospective cohort. Communities of targeted populations with reported exposure. 501 couples recruited upon discontinuing contraception to become pregnant, 2005-2009. None. Fecundability odds ratios (FORs) and

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service