Skip to Content
Merck
All Photos(1)

Key Documents

Y0000117

Halofantrine hydrochloride

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

1,3-Dichloro-a-[2-(dibutylamino)ethyl]-6-(trifluoromethyl)-9-phenanthrenemethanol hydrochloride, Halfan

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C26H30Cl2F3NO · HCl
CAS Number:
Molecular Weight:
536.88
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24
Pricing and availability is not currently available.

grade

pharmaceutical primary standard

API family

halofantrine

manufacturer/tradename

EDQM

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

Cl.CCCCN(CCCC)CCC(O)c1cc2c(Cl)cc(Cl)cc2c3cc(ccc13)C(F)(F)F

InChI

1S/C26H30Cl2F3NO.ClH/c1-3-5-10-32(11-6-4-2)12-9-25(33)23-16-22-21(14-18(27)15-24(22)28)20-13-17(26(29,30)31)7-8-19(20)23;/h7-8,13-16,25,33H,3-6,9-12H2,1-2H3;1H

InChI key

WANGFTDWOFGECH-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the Issuing Pharmacopoeia. For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Halofantrine hydrochloride EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Biochem/physiol Actions

Halofantrine is a blocker of delayed rectifier potassium current via the inhibition of hERG channel.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

related product

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Chronic 4

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jean-Yves Siriez et al.
Pathogens and global health, 106(2), 124-125 (2012-09-05)
In order to assess cardiac tolerance of halofantrine in children, we studied, retrospectively, 15 non complicated falciparum malaria cases treated with halofantrine, and focused on the effect on ventricular repolarisation. Our data showed that halofantrine can produce a moderate QTc
René Holm et al.
European journal of pharmaceutical sciences : official journal of the European Federation for Pharmaceutical Sciences, 42(4), 416-422 (2011-01-25)
The applicability of the semi-fluorinated alkane 1-perfluorohexyloctane (F6H8) as a novel excipient in lipid based drug delivery systems was studied. Solubility studies of 11 poorly water soluble drugs (cinnarizine, danazol, estradiol, fenofibrate, griseofulvin, halofantrine, lidocaine, prednisolone, probucol, rolipram and siramesine)
Kerenaftali Klein et al.
The Journal of pharmacy and pharmacology, 64(11), 1603-1613 (2012-10-13)
To investigate the population pharmacokinetics of the antimalarial halofantrine (HF) in healthy volunteers and patients with symptomatic falciparum malaria. Healthy volunteer data were obtained from six volunteers who received three different doses of HF (250, 500 and 1000 mg) after
Insaf F Khalil et al.
Journal of pharmaceutical and biomedical analysis, 54(1), 168-172 (2010-09-14)
Artemether-lumefantrine (ARM-LUM) has in recent years become the first-line treatment for uncomplicated malaria in many Sub-Saharan African countries. Vigorous monitoring of the therapeutic efficacy of this treatment is needed. This requires high-quality studies following standard protocols; ideally, such studies should
Jigar P Patel et al.
Chirality, 24(7), 558-565 (2012-05-17)
Experimental hyperlipidemia has shown to decrease cytochrome P450 3A4 and 2C11 expression and to increase liver concentrations and the plasma protein binding of halofantrine (HF) enantiomers. The present study examined the effect of hyperlipidemic (HL) serum on the metabolism of

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service