![Halofantrine hydrochloride European Pharmacopoeia (EP) Reference Standard](/deepweb/assets/sigmaaldrich/product/structures/323/009/e4f4b405-be49-4110-9108-2168bc5268a1/640/e4f4b405-be49-4110-9108-2168bc5268a1.png)
Halofantrine hydrochloride
![Halofantrine hydrochloride European Pharmacopoeia (EP) Reference Standard](/deepweb/assets/sigmaaldrich/product/structures/323/009/e4f4b405-be49-4110-9108-2168bc5268a1/640/e4f4b405-be49-4110-9108-2168bc5268a1.png)
Synonym(s):
1,3-Dichloro-a-[2-(dibutylamino)ethyl]-6-(trifluoromethyl)-9-phenanthrenemethanol hydrochloride, Halfan
Empirical Formula (Hill Notation):
C26H30Cl2F3NO · HCl
CAS Number:
Molecular Weight:
536.88
Products
Product Category
Brand
Color
Formula Weight
Manufacturer
Physical Form
Purity
Format
Quality Segment
Application
API Family
Special Grade
Showing 1-2 of 2
Compare
Product Number
Description
Pricing