Skip to Content
Merck
All Photos(1)

Documents

H9414

Sigma-Aldrich

Halofantrine hydrochloride

≥98% (HPLC), solid

Synonym(s):

1,3-Dichloro-a-[2-(dibutylamino)ethyl]-6-(trifluoromethyl)-9-phenanthrenemethanol hydrochloride, Halfan

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C26H30Cl2F3NO · HCl
CAS Number:
Molecular Weight:
536.88
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Assay

≥98% (HPLC)

form

solid

storage condition

desiccated

color

white to off-white

solubility

DMSO: >10 mg/mL

originator

GlaxoSmithKline

storage temp.

2-8°C

SMILES string

Cl.CCCCN(CCCC)CCC(O)c1cc2c(Cl)cc(Cl)cc2c3cc(ccc13)C(F)(F)F

InChI

1S/C26H30Cl2F3NO.ClH/c1-3-5-10-32(11-6-4-2)12-9-25(33)23-16-22-21(14-18(27)15-24(22)28)20-13-17(26(29,30)31)7-8-19(20)23;/h7-8,13-16,25,33H,3-6,9-12H2,1-2H3;1H

InChI key

WANGFTDWOFGECH-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

Halofantrine is a blocker of delayed rectifier potassium current via the inhibition of hERG channel.

Features and Benefits

This compound is featured on the Potassium Channels page of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.
This compound was developed by GlaxoSmithKline. To browse the list of other pharma-developed compounds and Approved Drugs/Drug Candidates, click here.

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Chronic 4

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

H Tie et al.
British journal of pharmacology, 130(8), 1967-1975 (2000-08-22)
Halofantrine is a widely used antimalarial agent which has been associated with prolongation of the 'QT interval' of the electrocardiogram (ECG), torsades de pointes and sudden death. Whilst QT prolongation is consistent with halofantrine-induced increases in cardiac ventricular action potential
René Holm et al.
European journal of pharmaceutics and biopharmaceutics : official journal of Arbeitsgemeinschaft fur Pharmazeutische Verfahrenstechnik e.V, 81(2), 281-287 (2012-04-03)
The bioavailability of the poorly soluble model drug halofantrine, dosed in a soy bean oil solution or in a self-nanoemulsifying drug delivery system (SNEDDS), at two levels of lipid, was assessed in rats. Three rat models were used: intact rats
Jigar P Patel et al.
Chirality, 24(7), 558-565 (2012-05-17)
Experimental hyperlipidemia has shown to decrease cytochrome P450 3A4 and 2C11 expression and to increase liver concentrations and the plasma protein binding of halofantrine (HF) enantiomers. The present study examined the effect of hyperlipidemic (HL) serum on the metabolism of
Martin Traebert et al.
European journal of pharmacology, 484(1), 41-48 (2004-01-20)
Several antimalarial drugs are known to produce a QT interval prolongation via a blockade of the rapidly activating delayed rectifier K+ current (IKr), encoded by the human-ether-a-go-go-related gene (hERG). We investigated the influence of lumefantrine and its major metabolite desbutyl-lumefantrine
Daria Van Tyne et al.
PLoS genetics, 7(4), e1001383-e1001383 (2011-05-03)
The Plasmodium falciparum parasite's ability to adapt to environmental pressures, such as the human immune system and antimalarial drugs, makes malaria an enduring burden to public health. Understanding the genetic basis of these adaptations is critical to intervening successfully against

Articles

We offer many products related to potassium channels for your research needs.

We offer many products related to potassium channels for your research needs.

We offer many products related to potassium channels for your research needs.

We offer many products related to potassium channels for your research needs.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service