Skip to Content
Merck
All Photos(3)

Documents

R5645

Sigma-Aldrich

Resorcinol

BioXtra, ≥99%

Synonym(s):

1,3-Benzenediol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H4-1,3-(OH)2
CAS Number:
Molecular Weight:
110.11
Beilstein:
906905
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.31

vapor density

3.8 (vs air)

Quality Level

vapor pressure

1 mmHg ( 21.1 °C)

product line

BioXtra

Assay

≥99%

form

powder

autoignition temp.

1126 °F

impurities

≤0.005% Phosphorus (P)
≤0.1% Insoluble matter

ign. residue

≤0.1%

bp

178 °C/16 mmHg (lit.)

mp

109-112 °C (lit.)

solubility

H2O: 1 M at 20 °C, clear, faintly yellow

anion traces

chloride (Cl-): ≤0.05%
sulfate (SO42-): ≤0.05%

cation traces

Al: ≤0.0005%
Ca: ≤0.0005%
Cu: ≤0.0005%
Fe: ≤0.0005%
K: ≤0.005%
Mg: ≤0.0005%
NH4+: ≤0.05%
Na: ≤0.005%
Pb: ≤0.001%
Zn: ≤0.0005%

SMILES string

Oc1cccc(O)c1

InChI

1S/C6H6O2/c7-5-2-1-3-6(8)4-5/h1-4,7-8H

InChI key

GHMLBKRAJCXXBS-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 3 - Eye Dam. 1 - Skin Irrit. 2 - Skin Sens. 1B - STOT SE 1 Oral - STOT SE 2 Oral

Target Organs

Central nervous system,Blood, Respiratory system

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 2

Flash Point(F)

260.6 °F - closed cup

Flash Point(C)

127 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Antonio B Fuertes et al.
Chemical communications (Cambridge, England), 48(49), 6124-6126 (2012-05-15)
Core@shell spheres made up of a thin layer of resorcinol-formaldehyde enveloping a silica core were prepared by means of a one-step method under Stöber conditions. These spheres are used as a platform for the synthesis of carbon or polymeric capsules
Phosphate-functionalized carbon monoliths from deep eutectic solvents and their use as monolithic electrodes in supercapacitors.
Daniel Carriazo et al.
ChemSusChem, 5(8), 1405-1409 (2012-07-06)
Anton V Gulevich et al.
Journal of the American Chemical Society, 134(12), 5528-5531 (2012-03-15)
The efficient Pd-catalyzed double-fold C-H oxygenation of arenes into resorcinols using the newly developed 2-pyrimidyldiisopropylsilyl (PyrDipSi) directing group is described. Its use allows for the sequential introduction of OAc and OPiv groups in a one-pot manner to produce orthogonally protected
S L da Silva et al.
European journal of medicinal chemistry, 44(1), 312-321 (2008-04-22)
Phospholipases A(2) are enzymes responsible for the hydrolysis of membrane phospholipids that release arachidonic acid, which serves as substrate for pro-inflammatory mediators, such as prostaglandins and leucotriens. The design of specific inhibitors for PLA(2) might help in the development of
Jinjun Li et al.
Environmental science & technology, 46(22), 12648-12654 (2012-10-25)
Graphitized carbons with mesoporous and macroporous structures were synthesized by a facile template-catalysis procedure using resorcinol and formaldehyde as carbon precursors and particulate hydrated metal oxides as both template and catalyst precursors. The materials were used as novel adsorbents for

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service