Skip to Content
Merck
All Photos(1)

Documents

W358908

Sigma-Aldrich

Resorcinol

≥98%, FG

Synonym(s):

1,3-Benzenediol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H4-1,3-(OH)2
CAS Number:
Molecular Weight:
110.11
FEMA Number:
3589
Beilstein:
906905
EC Number:
Council of Europe no.:
11250
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
4.047
NACRES:
NA.21

biological source

synthetic

grade

FG
Fragrance grade
Halal

Agency

follows IFRA guidelines
meets purity specifications of JECFA

reg. compliance

EU Regulation 1223/2009
EU Regulation 1334/2008 & 872/2012
FDA 21 CFR 110

vapor density

3.8 (vs air)

vapor pressure

1 mmHg ( 21.1 °C)

Assay

≥98%

form

flakes

autoignition temp.

1126 °F

bp

178 °C/16 mmHg (lit.)
178 °C/16 mmHg

mp

109-112 °C (lit.)

solubility

H2O: 5%, colorless to faintly yellow

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

fragrance allergen

no known allergens

Organoleptic

creamy; musty

SMILES string

Oc1cccc(O)c1

InChI

1S/C6H6O2/c7-5-2-1-3-6(8)4-5/h1-4,7-8H

InChI key

GHMLBKRAJCXXBS-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 3 - Eye Dam. 1 - Skin Irrit. 2 - Skin Sens. 1B - STOT SE 1 Oral - STOT SE 2 Oral

Target Organs

Central nervous system,Blood, Respiratory system

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 2

Flash Point(F)

260.6 °F - closed cup

Flash Point(C)

127 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Phosphate-functionalized carbon monoliths from deep eutectic solvents and their use as monolithic electrodes in supercapacitors.
Daniel Carriazo et al.
ChemSusChem, 5(8), 1405-1409 (2012-07-06)
Antonio B Fuertes et al.
Chemical communications (Cambridge, England), 48(49), 6124-6126 (2012-05-15)
Core@shell spheres made up of a thin layer of resorcinol-formaldehyde enveloping a silica core were prepared by means of a one-step method under Stöber conditions. These spheres are used as a platform for the synthesis of carbon or polymeric capsules
Jinjun Li et al.
Environmental science & technology, 46(22), 12648-12654 (2012-10-25)
Graphitized carbons with mesoporous and macroporous structures were synthesized by a facile template-catalysis procedure using resorcinol and formaldehyde as carbon precursors and particulate hydrated metal oxides as both template and catalyst precursors. The materials were used as novel adsorbents for
Ahmed M Elkhatat et al.
Advanced materials (Deerfield Beach, Fla.), 23(26), 2887-2903 (2011-05-25)
An overview on the preparation and properties of resorcinol-formaldehyde (RF) organic and carbon gels reveals the fascinating and remarkably flexible properties of RF carbon and organic gels and how these properties are related to the synthesis and processing conditions. The
Carsten Goebel et al.
Regulatory toxicology and pharmacology : RTP, 64(3), 459-465 (2012-10-17)
With the availability of the local lymph node assay, and the ability to evaluate effectively the relative skin sensitizing potency of contact allergens, a model for quantitative-risk-assessment (QRA) has been developed. This QRA process comprises: (a) determination of a no-expected-sensitisation-induction-level

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service