Skip to Content
Merck
All Photos(1)

Documents

PHR1281

Supelco

Albendazole

Pharmaceutical Secondary Standard; Certified Reference Material

Synonym(s):

Methyl 5-(propylthio)-2-benzimidazolecarbamate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H15N3O2S
CAS Number:
Molecular Weight:
265.33
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

certified reference material
pharmaceutical secondary standard

Quality Level

Agency

traceable to BP 763
traceable to Ph. Eur. A0325100
traceable to USP 1012553

API family

albendazole

CofA

current certificate can be downloaded

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-30°C

SMILES string

CCCSc1ccc2[nH]c(NC(=O)OC)nc2c1

InChI

1S/C12H15N3O2S/c1-3-6-18-8-4-5-9-10(7-8)14-11(13-9)15-12(16)17-2/h4-5,7H,3,6H2,1-2H3,(H2,13,14,15,16)

InChI key

HXHWSAZORRCQMX-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Pharmaceutical secondary standards for application in quality control, provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards. Albendazole (ABZ) is an anthelminthic drug, used in both human and veterinary medication. it is widely used to eliminated the effects of lymphatic filariasis.

Application

These Secondary Standards are qualified as Certified Reference Materials. These are suitable for use in several analytical applications including but not limited to pharma release testing, pharma method development for qualitative and quantitative analyses, food and beverage quality control testing, and other calibration requirements.

Analysis Note

These secondary standards offer multi-traceability to the USP, EP (PhEur) and BP primary standards, where they are available.

Other Notes

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Footnote

To see an example of a Certificate of Analysis for this material enter LRAA0016 in the slot below. This is an example certificate only and may not be the lot that you receive.

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Pictograms

Health hazardEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Repr. 2 - STOT RE 2 Oral

Target Organs

Adrenal gland,Blood,male reproductive organs,spleen

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

M E Oxberry et al.
Journal of veterinary pharmacology and therapeutics, 23(3), 113-120 (2000-12-08)
Trophozoites of the protozoan parasite Giardia duodenalis were exposed to various albendazole concentrations for 4 h, washed, fixed and incubated with antibodies raised against albendazole and its two major metabolites albendazole sulphoxide and albendazole sulphone. Tubulin antibodies were also used.
Marlieke E A de Kraker et al.
Tropical medicine & international health : TM & IH, 11(5), 718-728 (2006-04-28)
To determine the efficacies of combinations of ivermectin or diethylcarbamazine and albendazole, which are recommended for use in mass treatment programmes against lymphatic filariasis. Review of published trends in microfilarial (mf) intensities after treatment with these combination therapies. By fitting
Peiling Yap et al.
The American journal of tropical medicine and hygiene, 89(1), 23-31 (2013-05-22)
Post-treatment soil-transmitted helminth re-infection patterns were studied as part of a randomized controlled trial among school-aged children from an ethnic minority group in Yunnan province, People's Republic of China. Children with a soil-transmitted helminth infection (N = 194) were randomly
Ruth Ann Baird et al.
Neurology, 80(15), 1424-1429 (2013-04-10)
To review the evidence base for different treatment strategies in intraparenchymal neurocysticercosis in adults and children. A literature search of Medline, EMBASE, LILACS, and the Cochrane Database from 1980 to 2008, updated in 2012, resulted in the identification of 10
Aprilianto E Wiria et al.
PloS one, 8(3), e57899-e57899 (2013-03-26)
Helminth infections are proposed to have immunomodulatory activities affecting health outcomes either detrimentally or beneficially. We evaluated the effects of albendazole treatment, every three months for 21 months, on STH, malarial parasitemia and allergy. A household-based cluster-randomized, double-blind, placebo-controlled trial

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service