Skip to Content
Merck
All Photos(1)

Key Documents

35032

Supelco

Fenbendazole

VETRANAL®, analytical standard

Synonym(s):

Methyl 5-(phenylthio)-2-benzimidazolecarbamate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H13N3O2S
CAS Number:
Molecular Weight:
299.35
Beilstein:
759077
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24
Pricing and availability is not currently available.

grade

analytical standard

Quality Level

product line

VETRANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

clinical testing

format

neat

SMILES string

COC(=O)Nc1nc2cc(Sc3ccccc3)ccc2[nH]1

InChI

1S/C15H13N3O2S/c1-20-15(19)18-14-16-12-8-7-11(9-13(12)17-14)21-10-5-3-2-4-6-10/h2-9H,1H3,(H2,16,17,18,19)

InChI key

HDDSHPAODJUKPD-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Chemical structure: benzimidazole
Fenbendazole is a thio substituted benzimidazole, which belongs to the group of anthelmintics.[1] It can be widely used in veterinary medicine particularly, in the treatment of helminth infections.[2]

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

VETRANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

related product

Product No.
Description
Pricing

Pictograms

Health hazardEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Repr. 2 - STOT RE 2

Target Organs

Liver,lymph node,Stomach,Nervous system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Determination of fenbendazole and oxfendazole in liver and muscle using liquid chromatography?mass spectrometry
Blanchflower J.W, et al.
Analyst, 119, 1325-1328 (1994)
Interaction between fenbendazole and piperonyl butoxide: pharmacokinetic and pharmacodynamic implications
Benchaoui.A.H and Mckellar.A.Q
The Journal of Pharmacy and Pharmacology, 48, 753-759 (1996)
Aileen Adam et al.
Veterinary parasitology, 177(3-4), 383-386 (2011-01-18)
On red grouse estates in the UK the nematode parasite Trichostrongylus tenuis is often controlled by application of grit medicated with the anthelmintic fenbendazole (FBZ). To date, assessment of the efficacy has been inhibited by the inability to quantify uptake
Moreno Rossi et al.
Journal of feline medicine and surgery, 13(10), 793-795 (2011-08-13)
An 8-month-old owned European cat showing abdominal pain, fever, distended painful bladder and urinary blockage was presented. Intravenous fluids were immediately administered and, after sedation, a urinary catheter was applied. Blood and urine analysis revealed cystitis and a moderate-to-severe degree
A C Clark et al.
The Journal of small animal practice, 54(2), 99-103 (2012-11-14)
A 4-year-old, spayed-female great Dane was referred for surgical treatment of a suspected meningioma, diagnosed on magnetic resonance imaging 10 days prior to presentation. The suspected meningioma was removed via image-guided stereotactic craniotomy. Histopathological diagnosis was severe, locally extensive, chronic meningoencephalitis

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service