The 5% MPA solution should be made without considering purity (i.e. 5 g of cat#239275 in 100 g of water). Please note that the 5% MPA should be prepared fresh each day it is to be used.
MAK440
Glutathione GSH/GSSG Assay Kit
Sufficient for 100 colorimetric tests
Synonym(s):
GSH/GSSG Ratio Kit
About This Item
Recommended Products
Quality Level
application(s)
pharmaceutical
detection method
colorimetric
relevant disease(s)
Alzheimer′s disease; Parkinson′s disease; cancer; neurological disorders; aging/geriatric diseases
storage temp.
−20°C
Related Categories
General description
Application
- Cancer Research
- Diabetes Research
- Neurodegenerative Disease Research
Features and Benefits
Simplified Process: Experience a streamlined process with the addition of only a single working reagent and a 10 minute room temperature reaction, reducing complexity and saving valuable time and effort.
Compatibility with High-Throughput Systems: Easily incorporate our kit into high-throughput handling systems, ensuring smooth and accurate processing, enhancing efficiency in your laboratory workflow.
Suitability
Principle
Other Notes
Storage Class Code
10 - Combustible liquids
Flash Point(F)
188.6 °F
Flash Point(C)
87 °C
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
-
How to make 5% MPA? The MPA recommended in the manual (239275) is 35% pure. Does 5%(w/v) refer to the concentration of pure MPA or the impure solid mixture? In this case do I need to account for the 35%?
1 answer-
Helpful?
-
-
How can I determine the shelf life / expiration / retest date of this product?
1 answer-
If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdfHelpful?
-
-
How is shipping temperature determined? And how is it related to the product storage temperature?
1 answer-
Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf
Helpful?
-
-
Hi, Can I use this kit for testing GSH/GSSG ratio in plants (Arabidopsis), if yes, is there any modifications in the protocol for plants? Thank you
1 answer-
This kit can work on whole blood, plasma, serum, urine, tissue, and cell extracts samples; however, it has not been tested for plant samples, including Arabidopsis. Therefore, no specific modification in the protocol is made to test the GSH/GSSG ratio in plants using this kit.
Helpful?
-
-
After the extraction procedure on colon tissues, do I have to also realize the deproteination before the assay ?
1 answer-
Colon tissue samples must undergo the deproteination procedure as described on page 2 of the kit's technical bulletin.
Helpful?
-
-
Can this kit be used to analyze plasma or serum samples? If yes, is plasma or serum better? If yes, what is the procedure to follow as the procedure is described for whole blood and cell lysate only. What is the stability of frozen plasma for glutathione?
1 answer-
Serum or plasma samples can be prepared using the same method as whole blood samples. Both types of samples are suitable for testing and are equally recommended. EDTA plasma is specifically known to be compatible with the assay.
Samples of serum or plasma can be frozen for later analysis; however, it is advisable to freeze them promptly after preparation at -80°C. To minimize the risk of glutathione degradation, avoid multiple freeze-thaw cycles and aim to test the samples within one month.
Helpful?
-
-
Where does the DF of the calculation come from when using Scavenger or without Scavenger? If I need to make an additional dilution do I multiply it?
1 answer-
The volumes used for buffer and scavenger should not be altered from the stated volumes.
For all samples treated with Scavenger, DF = 165. For samples not treated with Scavenger, DF = 450 for whole blood and 150 for all other samples. Please refer to the technical bulletin, specifically page 4, for information on the calculation of GSH and GSSG.Helpful?
-
-
How is the test for GSSG and total GSH performed when using dry biomass? Is only PBS used or should phosphate buffer + EDTA be added? What is the estimated sonication time?
1 answer-
In this case, the use of PBS is not recommended. For preparation of a sample from the dry biomass, follow the procedures as described for preparing a cell lysate. That is, use 50 mM phosphate, pH 7, 1 mM EDTA, and 20 microliters of Scavenger for preparation of the sample for GSSG measurement, and use 50 mM phosphate, pH 7, and 1 mM EDTA (with NO scavenger) for preparation of the sample for GSH measurement. Sonication for 30-60 seconds should be sufficient, but examination of a sample under a microscope to ensure that it is completely homogenized is recommended.
Helpful?
-
-
To which type of tissues this kit is suitable for? was it tested on mice brain?
1 answer-
This product is not tested specifically on mouse brain tissue. However, this kit is very robust and suitable for use on all species. Please see the link below to review an article referencing the use of this item on mouse brain:
Thiosulfate sulfurtransferase deficiency promotes oxidative distress and aberrant NRF2 function in the brain
https://www.ncbi.nlm.nih.gov/pmc/articles/PMC10701433/Helpful?
-
-
We want to buy the glutathione Assay Kit to detect the GSH/GSSG level in bile sample and we also found one publication(PMID: 23978715) used the product from your company. However, I can't find how to prepare the bile sample in the product manual.
1 answer-
This kit has not been tested for use with bile. Please see the link below to review a publication that indicates suitability: https://www.ncbi.nlm.nih.gov/pmc/articles/PMC3362073/
Helpful?
-
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service