Skip to Content
Merck
All Photos(7)

Key Documents

P5631

Sigma-Aldrich

o-Cresolphthalein Complexone

powder

Synonym(s):

o-Cresolphthalein-3′,3′′-bis-methyleneiminodiacetic acid, o-Cresolphthalexon, Metal phthalein, Phthalein purple, Xylenylphthalein-bisiminodiacetic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C32H32N2O12
CAS Number:
Molecular Weight:
636.60
Beilstein:
381994
EC Number:
MDL number:
UNSPSC Code:
12171500
PubChem Substance ID:
NACRES:
NA.47

form

powder

Quality Level

technique(s)

titration: suitable

color

white to off-white

mp

181-185 °C (dec.) (lit.)

solubility

methanol: 1 mg/mL

suitability

suitable for determination of calcium

application(s)

diagnostic assay manufacturing
hematology
histology

storage temp.

room temp

SMILES string

Cc1cc(cc(CN(CC(O)=O)CC(O)=O)c1O)C2(OC(=O)c3ccccc23)c4cc(C)c(O)c(CN(CC(O)=O)CC(O)=O)c4

InChI

1S/C32H32N2O12/c1-17-7-21(9-19(29(17)43)11-33(13-25(35)36)14-26(37)38)32(24-6-4-3-5-23(24)31(45)46-32)22-8-18(2)30(44)20(10-22)12-34(15-27(39)40)16-28(41)42/h3-10,43-44H,11-16H2,1-2H3,(H,35,36)(H,37,38)(H,39,40)(H,41,42)

InChI key

IYZPEGVSBUNMBE-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

o-Cresolphthalein Complexone is a pH indicator dye commonly used for the calorimetric determination of calcium in serum.

Application

o-Cresolphthalein Complexone has been used in the estimation of calcium in serum samples based on the reaction of Ca2+ with o-cresolphthalein complexone in an alkaline solution.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

C M Corns et al.
Annals of clinical biochemistry, 24 ( Pt 4), 345-351 (1987-07-01)
The reasons for the non-linearity of the standard curve for the calcium-cresolphthalein complexone reaction have been investigated. Calcium forms both 1:1 and 2:1 complexes with cresolphthalein complexone; at low calcium concentrations the 1:1 complex predominates and causes non-linearity. At high
J Toffaletti et al.
Clinical chemistry, 26(11), 1562-1565 (1980-10-01)
We adapted a method for dialyzable calcium from fluorometric detection by use of calcein to a more specific spectrophotometric determination with cresolphthalein complexone. The reagents are available commercially and perform satisfactorily with respect to noise level, drift, stability, and sensitivity.
James H Wible et al.
Radiology, 233(1), 158-164 (2004-09-30)
To measure serum calcium concentration with three different analytic methods after administration of gadoversetamide and three other gadolinium chelates in dogs. Six dogs were injected with 0.1-, 0.3-, 1.0-, and 3.0-mmol/kg doses of gadoversetamide; 1.0-mmol/kg doses of gadodiamide, gadopentetate, and
C M Corns
Annals of clinical biochemistry, 27 ( Pt 2), 152-155 (1990-03-01)
The spectral interference caused by haemoglobin in the calcium-cresolphthalein complexone reaction was investigated. The results show that the nature of the haemoglobin derivative in the reaction mixture influences the degree of interference; in turn, the derivative formed depends on the
B Aghabeigi et al.
International journal of oral and maxillofacial surgery, 31(6), 646-649 (2003-01-11)
Saline aspirates have been commonly used in the biochemical investigations of temporomandibular joint (TMJ) pathology. However, due to presence of adhesions in the diseased temporomandibular joint, full equilibration between the injected saline and the synovial fluid may not be achieved

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service