Skip to Content
Merck
All Photos(1)

Key Documents

08581

Sigma-Aldrich

2-Amino-2-methyl-1-propanol

technical, ≥90% (GC)

Synonym(s):

β-Aminoisobutyl alcohol, AMP

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)2C(NH2)CH2OH
CAS Number:
Molecular Weight:
89.14
Beilstein:
505979
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

vapor density

3 (vs air)

Quality Level

vapor pressure

<1 mmHg ( 25 °C)

grade

technical

Assay

≥90% (GC)

form

solid

refractive index

n20/D 1.4455 (lit.)

useful pH range

9.0-10.5

pKa (25 °C)

9.7

bp

165 °C (lit.)

mp

24-28 °C (lit.)

density

0.934 g/mL at 25 °C (lit.)

functional group

amine
hydroxyl

SMILES string

CC(C)(N)CO

InChI

1S/C4H11NO/c1-4(2,5)3-6/h6H,3,5H2,1-2H3

InChI key

CBTVGIZVANVGBH-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

Application

2-Amino-2-methyl-1-propanol (AMP) is a sterically hindered amine that can be used:
  • As an acid gas treating solvent to remove CO2 and H2S from gas streams by absorption.
  • As a reagent in the synthesis of a fluorophore, silicon-rhodamine dye, for bioimaging.

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Chronic 3 - Eye Dam. 1 - Skin Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

179.8 °F - closed cup

Flash Point(C)

82.1 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Characterization and comparison of the CO2 absorption performance into single and blended alkanolamines in a packed column.
Aroonwilas A and Veawab A
Industrial & Engineering Chemistry Research, 43(9), 2228-2237 (2004)
A near-infrared fluorophore for live-cell super-resolution microscopy of cellular proteins.
Lukinavicius, G, et al.
Nature Chemistry, 5(2), 132-132 (2013)
Selective absorption of H2S from gas streams containing H2S and CO2 into aqueous solutions of N-methyldiethanolamine and 2-amino-2-methyl-1-propanol.
Mandal BP, et al.
Separation and Purification Technology, 35(3), 191-202 (2004)
Modeling of CO2 capture by three typical amine solutions in hollow fiber membrane contactors.
Wang R, et al.
Chemical Engineering and Processing, 43(7), 849-856 (2004)
Pei Zhang et al.
Journal of environmental sciences (China), 20(1), 39-44 (2008-06-25)
To improve the efficiency of the carbon dioxide cycling process and to reduce the regeneration energy consumption, a sterically hindered amine of 2-amino-2-methyl-1-propranol (AMP) was investigated to determine its regeneration behavior as a CO2 absorbent. The CO2 absorption and amine

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service