Skip to Content
Merck

D2376

Deoxyribonucleic acid methyl green

DNAse substrate

Synonym(s):

Deoxyribonucleic acid–methyl green complex, DNA-Methyl Green

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

UNSPSC Code:
12352204
NACRES:
NA.32
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

form

solid

storage temp.

−20°C

General description

Substrate for DNase
Methyl green is a widely used nuclei counterstain for histochemical methods. Methyl green binds to the DNA major groove and is used for tracking triplex formation. The positive charge on the aniline rings in the methyl green enables its strong binding to DNA. Methyl green aids clear visualization in confocal laser scanning microscopy.

Application

Deoxyribonucleic acid methyl green has been used in DNA methyl green bioassay.


flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)

Hazard Classifications

Aquatic Chronic 3 - Eye Dam. 1 - Met. Corr. 1 - Skin Corr. 1A

Storage Class

8A - Combustible corrosive hazardous materials

pictograms

Corrosion

signalword

Danger



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library