Skip to Content
Merck

A0638

Adipic acid dihydrazide

≥98% (titration)

Synonym(s):

Hexanedihydrazide

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
NH2NHCO(CH2)4CONHNH2
CAS Number:
Molecular Weight:
174.20
UNSPSC Code:
12352127
NACRES:
NA.25
PubChem Substance ID:
EC Number:
213-999-5
Beilstein/REAXYS Number:
973863
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Level

assay

≥98% (titration)

form

powder

reaction suitability

reagent type: cross-linking reagent

mp

180-182 °C (lit.)

solubility

H2O: 100 mg/mL

storage temp.

−20°C

SMILES string

NNC(=O)CCCCC(=O)NN

InChI

1S/C6H14N4O2/c7-9-5(11)3-1-2-4-6(12)10-8/h1-4,7-8H2,(H,9,11)(H,10,12)

InChI key

IBVAQQYNSHJXBV-UHFFFAOYSA-N

General description

Adipic acid dihydrazide is a homobifunctional cross-linking reagent that is specific for aldehydes. This results in relatively stable hydrazone linkages. Adipic acid dihydrazide is generally used in the linking of glycoproteins, like antibodies, in a site-specific fashion following periodate oxidation. Oxidation and coupling may be performed at pH 5.0 due to the low pKa of the hydrazide which avoids competition by primary amines.

Application

Adipic acid dihydrazide has been used:
  • in the preparation of reactive premix to synthesize the porous biomaterial
  • for the crosslinking of methacrylated chondroitin sulfate (MA-CS) coating using carbodiimide-based chemistry for the production and characterization of methacrylated chondroitin sulfate magnetic nanoparticles (MA-CS MNPs)
  • for the covalent labeling of rhamnolipids, pyochelin, and vancomycin with Abberior STARNHS ester dye

Biochem/physiol Actions

Adipic acid dihydrazide (ADH) is a low molecular weight compound that comprises a hydrazide group at each end. This leads to the supply of extra adsorption sites for heavy metals that maintain or elevate the adsorption capacities of the cross-linked adsorbents. ADH is used as a crosslinker in various fields, like making mechanical latexes films and injectable oxidized hyaluronic acid hydrogel.

Other Notes

Note that the hydrazone bond may be stabilized by selective reduction using, for example, sodium cyanoborohydride.


Still not finding the right product?

Explore all of our products under Adipic acid dihydrazide


pictograms

Environment

hcodes

Hazard Classifications

Aquatic Chronic 2

Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

302.0 °F - closed cup

flash_point_c

150 °C - closed cup

ppe

Eyeshields, Gloves, type N95 (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Tulsidas G Shrivastav et al.
Journal of immunoassay & immunochemistry, 33(1), 1-17 (2011-12-21)
The introduction of spacers in coating steroid antigen or enzyme conjugates or immunogen is known to exert an influence on the sensitivity of steroid enzyme immunoassays. We have introduced different homobifunctional spacers having varying atomic length (3 to 10) between
Bartosz Gerard Gdaniec et al.
iScience, 25(1), 103669-103669 (2022-01-15)
Efficient delivery of toxic compounds to bacterial competitors is essential during interspecies microbial warfare. Rhamnolipids (RLPs) are glycolipids produced by Pseudomonas and Burkholderia species involved in solubilization and uptake of environmental aliphatic hydrocarbons and perform as biosurfactants for swarming motility.
Lin-Song Li et al.
Pharmaceuticals (Basel, Switzerland), 14(2) (2021-02-03)
Functionalized gold nanoparticles (AuNPs) have been successfully used in many fields as a result of having low cytotoxicity, good biocompatibility, excellent optical properties, and their ability to target cancer cells. Here, we synthesized AuNP carriers that were modified by hyaluronic



Global Trade Item Number

SKUGTIN
A0638-100G04061835326204
A0638-25G04061833334201