Skip to Content
Merck
All Photos(1)

Key Documents

M0600905

Metamizole impurity A

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

4-Formylaminoantipyrine, N-(1,5-Dimethyl-3-oxo-2-phenyl-2,3-dihydro-1H-pyrazol-4-yl)formamide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H13N3O2
CAS Number:
Molecular Weight:
231.25
Beilstein:
217665
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

metamizole

manufacturer/tradename

EDQM

application(s)

pharmaceutical (small molecule)

format

neat

SMILES string

CN1N(C(=O)C(NC=O)=C1C)c2ccccc2

InChI

1S/C12H13N3O2/c1-9-11(13-8-16)12(17)15(14(9)2)10-6-4-3-5-7-10/h3-8H,1-2H3,(H,13,16)

InChI key

WSJBSKRPKADYRQ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the Issuing Pharmacopoeia. For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Metamizole impurity A EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

S Imaoka et al.
Archives of biochemistry and biophysics, 265(1), 159-170 (1988-08-15)
Four aminopyrine metabolites generated by hepatic microsomes were simultaneously assayed by high-performance liquid chromatography. The metabolites were 4-monomethylaminoantipyrine (MAA), 4-aminoantipyrine (AA), 3-hydroxymethyl-2-methyl-4-dimethylamino-1-phenyl-3-pyrazoline-5-one (AM-OH), and one unidentified metabolite. MAA was the major metabolite generated by the microsomes; its formation was induced
K Matsuyama et al.
Journal of pharmacobio-dynamics, 6(11), 821-828 (1983-11-01)
The effect of phenobarbital (PB) and 3-methylcholanthrene (3-MC) on the metabolic behavior of aminopyrine (AM) was studied using an isolated hepatocyte system prepared from male Wistar rats. The formation of 4-formylaminoantipyrine (FAA) was increased after pretreatment with PB, but not
K Inoue et al.
Journal of chromatography, 274, 201-208 (1983-05-13)
Aminopyrine and its metabolites, including 3-hydroxymethyl-2-methyl-4-dimethylamino-1-phenyl-3-pyrazoline-5-one which is a hydroxylated metabolite of aminopyrine, were separated on a reversed-phase (C8) Radial-Pak column using a mobile phase of methanol-triethylamine-water (30:1:69) adjusted to pH 5.40 with acetic acid. Detection of the peak was
I Carretero et al.
The Analyst, 120(6), 1729-1732 (1995-06-01)
A rapid solid-phase extraction (SPE) procedure was developed for the quantitative isolation of three important antipyrine (dipyrone) metabolites from human plasma: 4-formylaminoantipyrine (FAA), 4-aminoantipyrine (AA) and 4-methylaminoantipyrine (MAA). Separation and quantitation were performed using micellar liquid chromatography (MLC) with a
High-performance liquid chromatographic analysis of dipyrone metabolites to study their formation in human liver microsomes.
G Geisslinger et al.
Pharmaceutical research, 13(8), 1272-1275 (1996-08-01)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service