Skip to Content
Merck
All Photos(2)

Key Documents

32595

Supelco

4-Formylaminoantipyrine

VETRANAL®, analytical standard

Synonym(s):

N-(1,5-Dimethyl-3-oxo-2-phenyl-2,3-dihydro-1H-pyrazol-4-yl)formamide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H13N3O2
CAS Number:
Molecular Weight:
231.25
Beilstein:
217665
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

VETRANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

pharmaceutical

format

neat

SMILES string

CN1N(C(=O)C(NC=O)=C1C)c2ccccc2

InChI

1S/C12H13N3O2/c1-9-11(13-8-16)12(17)15(14(9)2)10-6-4-3-5-7-10/h3-8H,1-2H3,(H,13,16)

InChI key

WSJBSKRPKADYRQ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

4-Formylaminoantipyrines are grouped under the class of biodegradable organic micropollutants (OMPs).

Application

4-Formylaminoantipyrine may be used as an analytical reference standard for the determination of the analyte in:
  • Aqueous samples by ultra-high performance liquid chromatography-high resolution mass spectrometry (UHPLC-HRMS).
  • Groundwater, surface water and wastewater samples by online mixed-bed multilayer solid-phase extraction (SPE) and HPLC combined with tandem mass spectrometry (MS/MS).

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

VETRANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Multiresidue analytical method for the simultaneous determination of 72 micropollutants in aqueous samples with ultra high performance liquid chromatography-high resolution mass spectrometry.
Wode F, et al.
Journal of Chromatography A, 1270, 118-126 (2012)
Integrating organic micropollutant removal into tertiary filtration: Combining PAC adsorption with advanced phosphorus removal.
Altmann J, et al.
Water Research, 84, 58-65 (2015)
K Inoue et al.
Journal of chromatography, 274, 201-208 (1983-05-13)
Aminopyrine and its metabolites, including 3-hydroxymethyl-2-methyl-4-dimethylamino-1-phenyl-3-pyrazoline-5-one which is a hydroxylated metabolite of aminopyrine, were separated on a reversed-phase (C8) Radial-Pak column using a mobile phase of methanol-triethylamine-water (30:1:69) adjusted to pH 5.40 with acetic acid. Detection of the peak was
High-performance liquid chromatographic analysis of dipyrone metabolites to study their formation in human liver microsomes.
G Geisslinger et al.
Pharmaceutical research, 13(8), 1272-1275 (1996-08-01)
I Carretero et al.
The Analyst, 120(6), 1729-1732 (1995-06-01)
A rapid solid-phase extraction (SPE) procedure was developed for the quantitative isolation of three important antipyrine (dipyrone) metabolites from human plasma: 4-formylaminoantipyrine (FAA), 4-aminoantipyrine (AA) and 4-methylaminoantipyrine (MAA). Separation and quantitation were performed using micellar liquid chromatography (MLC) with a

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service