Skip to Content
Merck
All Photos(1)

Key Documents

M0600900

Metamizole sodium

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

Dipyrone monohydrate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C13H16N3NaO4S · H2O
CAS Number:
Molecular Weight:
351.35
UNSPSC Code:
41116107
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

metamizole

manufacturer/tradename

EDQM

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

InChI

1S/C13H17N3O4S.Na.H2O/c1-10-12(14(2)9-21(18,19)20)13(17)16(15(10)3)11-7-5-4-6-8-11;;/h4-8H,9H2,1-3H3,(H,18,19,20);;1H2/q;+1;/p-1

InChI key

UNZIDPIPYUMVPA-UHFFFAOYSA-M

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the Issuing Pharmacopoeia. For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Metamizole sodium EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Pictograms

Health hazard

Signal Word

Warning

Hazard Statements

Hazard Classifications

Repr. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

C G Manole et al.
Journal of cellular and molecular medicine, 15(11), 2284-2296 (2011-09-08)
We used rat experimental myocardial infarction to study the ultrastructural recovery, especially neo-angiogenesis in the infarction border zone. We were interested in the possible role(s) of telocytes (TCs), a novel type of interstitial cell very recently discovered in myocardim (see
Update on the incidence of metamizole sodium-induced blood dyscrasias in Poland.
Basak G W, et al.
The Journal of International Medical Research, 38(4), 1374-1380 (2010)
Acute kidney injury associated with metamizole sodium ingestion.
Hassan K, et al.
Renal Failure, 33(5), 544-547 (2011)
E Marciniak et al.
Domestic animal endocrinology, 58, 97-103 (2016-10-30)
This study tested the hypothesis that salsolinol, a derivative of dopamine, affects GnRH and LH secretion in lactating sheep. In the in vivo experiment, the structural analogue of salsolinol, 1-methyl-3,4-dihydroisoquinoline (1-MeDIQ), was infused into the infundibular nucleus-median eminence of sheep at
Berenice Ovalle-Magallanes et al.
The Journal of pharmacy and pharmacology, 67(12), 1733-1743 (2015-09-27)
This work was undertaken to assess the antinociceptive and hypoglycaemic properties of a quantified extract of Conyza filaginoides (CFOE), as well as the antinociceptive potential of rutin, the main active compound of the plant, in normoglycaemic and/or hyperglycaemic mice (nicotinamide-streptozotocin

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service