Skip to Content
Merck
All Photos(1)

Key Documents

BP421

Ethinylestradiol

British Pharmacopoeia (BP) Assay Standard

Synonym(s):

17α-Ethynylestradiol, 17α-Ethynyl-1,3,5(10)-estratriene-3,17β-diol, 19-Nor-1,3,5(10),17α-pregnatrien-20-yne-3,17-diol, Ethinylestradiol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H24O2
CAS Number:
Molecular Weight:
296.40
Beilstein:
2419975
EC Number:
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24
Pricing and availability is not currently available.

grade

pharmaceutical primary standard

API family

ethinyl estradiol

form

solid

shelf life

limited shelf life, expiry date on the label

manufacturer/tradename

BP

mp

182-183 °C (lit.)

application(s)

pharmaceutical
pharmaceutical small molecule

format

neat

storage temp.

2-8°C

SMILES string

[H][C@]12CC[C@@]3(C)[C@@]([H])(CC[C@@]3(O)C#C)[C@]1([H])CCc4cc(O)ccc24

InChI

1S/C20H24O2/c1-3-20(22)11-9-18-17-6-4-13-12-14(21)5-7-15(13)16(17)8-10-19(18,20)2/h1,5,7,12,16-18,21-22H,4,6,8-11H2,2H3/t16-,17-,18+,19+,20+/m1/s1

InChI key

BFPYWIDHMRZLRN-SLHNCBLASA-N

Gene Information

human ... ESR1(2099)

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.
For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Ethinylestradiol BP Reference standard, intended for use in laboratory tests only as specifically prescribed in the British Pharmacopoeia.

Also used in monographs such as:
  • Co-cyprindiol Tablets
  • Levonorgestrel Tablets
  • Norgestrel Tablets
  • Levonorgestrel and Ethinylestradiol Tablets
  • Ethinylestradiol Tablets
  • Other Notes

    Sales restrictions may apply.

    Signal Word

    Danger

    Hazard Statements

    Hazard Classifications

    Acute Tox. 4 Oral - Aquatic Chronic 1 - Carc. 2 - Repr. 1A

    Storage Class Code

    6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

    WGK

    WGK 3


    Choose from one of the most recent versions:

    Certificates of Analysis (COA)

    Lot/Batch Number

    It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

    If you need assistance, please contact Customer Support.

    Already Own This Product?

    Find documentation for the products that you have recently purchased in the Document Library.

    Visit the Document Library

    Andrew M Kaunitz et al.
    Obstetrics and gynecology, 123(2 Pt 1), 295-303 (2014-01-10)
    To compare a new low-dose levonorgestrel and ethinyl estradiol contraceptive patch (Patch) with a combination oral contraceptive (Pill; 100 micrograms levonorgestrel, 20 micrograms ethinyl estradiol) regarding efficacy, safety, compliance, and unscheduled uterine bleeding. Women (17-40 years; body mass index 16-60)
    Matteo Morotti et al.
    European journal of obstetrics, gynecology, and reproductive biology, 183, 178-182 (2014-12-03)
    To evaluate the effect of a desogestrel progestogen-only pill (POP) compared to continuous combined oral contraception (COC) on migraine patterns in women with migraine without aura. A retrospective analysis of prospective headache charts from migrainous women who used the POP
    Marie Louise Wissing et al.
    Journal of ovarian research, 7, 110-110 (2014-11-30)
    Corona radiata cells (CRCs) refer to the fraction of cumulus cells just adjacent to the oocyte. The CRCs are closely connected to the oocyte throughout maturation and their gene expression profiles might reflect oocyte quality. Polycystic ovary syndrome (PCOS) is
    Christoph Kapitza et al.
    Journal of clinical pharmacology, 55(5), 497-504 (2014-12-06)
    The effect of semaglutide, a once-weekly human glucagon-like peptide-1 (GLP-1) analog in development for type 2 diabetes (T2D), on the bioavailability of a combined oral contraceptive was investigated. Postmenopausal women with T2D (n = 43) on diet/exercise ± metformin received ethinylestradiol (0.03 mg)/levonorgestrel (0.15 mg) once
    Gregory Brazzola et al.
    Evolutionary applications, 7(9), 1084-1093 (2015-01-02)
    The evolutionary potential of natural populations to adapt to anthropogenic threats critically depends on whether there exists additive genetic variation for tolerance to the threat. A major problem for water-dwelling organisms is chemical pollution, and among the most common pollutants

    Questions

    Reviews

    No rating value

    Active Filters

    Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

    Contact Technical Service