Skip to Content
Merck
All Photos(1)

Documents

B5274

Supelco

Bupivacaine hydrochloride monohydrate

analytical standard, for drug analysis

Synonym(s):

1-Butyl-N-(2,6-dimethylphenyl)-2-piperidinecarboxamide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H28N2O · HCl · H2O
CAS Number:
Molecular Weight:
342.90
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard, for drug analysis

Quality Level

Assay

≥99%

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

forensics and toxicology
pharmaceutical (small molecule)
veterinary

format

neat

SMILES string

O.Cl.CCCCN1CCCCC1C(=O)Nc2c(C)cccc2C

InChI

1S/C18H28N2O.ClH.H2O/c1-4-5-12-20-13-7-6-11-16(20)18(21)19-17-14(2)9-8-10-15(17)3;;/h8-10,16H,4-7,11-13H2,1-3H3,(H,19,21);1H;1H2

InChI key

HUCIWBPMHXGLFM-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Bupivacaine hydrochloride monohydrate may be used as an analytical standard for the determination of the bupivacaine in pharmaceutical preparations and plasma samples using chromatography techniques.
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Biochem/physiol Actions

Sodium channel blocker, local anesthetic.

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 1 Inhalation - Acute Tox. 2 Dermal - Acute Tox. 2 Oral

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Determination of free concentration of ropivacaine and bupivacaine in blood plasma by ultrafiltration and coupled-column liquid chromatography
Arvidsson T and Eklund E
Journal of Chromatography. B, Biomedical Sciences and Applications, 668(1), 91-98 (1995)
Arnaud Chaumeron et al.
Clinical orthopaedics and related research, 471(7), 2284-2295 (2013-03-22)
The postoperative analgesic potential of periarticular anesthetic infiltration (PAI) after TKA is unclear as are the complications of continuous femoral nerve block on quadriceps function. We asked (1) whether PAI provides equal or improved postoperative pain control in comparison to
Huiyuan Zhang et al.
Drug design, development and therapy, 13, 1659-1666 (2019-06-14)
Background: Bupivacaine (Bup) is the most commonly used local anesthetic. However, Bup induces cytotoxicity, especially in older patients. Recent reports have indicated that andrographolide (Andro) exhibits protective effects on human neurons. Nevertheless, whether Andro can inhibit Bup-induced cytotoxicity remains unclear.
Determination of local anaesthetics and their impurities in pharmaceutical preparations using HPLC method with amperometric detection
Fijalek Z, et al.
Journal of Pharmaceutical and Biomedical Analysis, 37(5), 913-918 (2005)
Kejian Shi et al.
Anesthesia and analgesia, 116(4), 804-809 (2013-03-06)
While lipid emulsion may reverse the systemic toxicity of bupivacaine, the pharmacokinetics and tissue distribution of bupivacaine after lipid emulsion infusion are not clear. In this study, we assessed the influence of lipid emulsion administration on the pharmacokinetics and tissue

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service