Skip to Content
Merck
All Photos(1)

Documents

L0600000

Lidocaine hydrochloride

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

Lidocaine hydrochloride monohydrate, 2-Diethylamino-N-(2,6-dimethylphenyl)acetamide hydrochloride monohydrate, Lignocaine hydrochloride monohydrate, Xylocaine hydrochloride monohydrate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H22N2O · HCl · H2O
CAS Number:
Molecular Weight:
288.81
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

lidocaine

manufacturer/tradename

EDQM

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

pharmaceutical (small molecule)

format

neat

SMILES string

Cl[H].[H]O[H].CCN(CC)CC(=O)Nc1c(C)cccc1C

InChI

1S/C14H22N2O.ClH.H2O/c1-5-16(6-2)10-13(17)15-14-11(3)8-7-9-12(14)4;;/h7-9H,5-6,10H2,1-4H3,(H,15,17);1H;1H2

InChI key

YECIFGHRMFEPJK-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the Issuing Pharmacopoeia. For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Lidocaine hydrochloride EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Biochem/physiol Actions

Na+ channel blocker; class IB antiarrhythmic that is rapidly absorbed after parenteral administration

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

related product

Product No.
Description
Pricing

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Shiran Katabi et al.
STAR protocols, 5(2), 103081-103081 (2024-05-25)
Extracellular recordings in behaving animals are useful for establishing associations between neuronal activity and behavior. Here, we describe how to record in the external globus pallidus (GPe) of monkeys engaged in a behavioral task. We detail the stereotaxic surgery for
Fang-Chi Yang et al.
Neurobiology of learning and memory, 112, 186-194 (2013-07-31)
Learning active or reactive responses to fear involves different brain circuitry. This study examined how the nuclus accumbens (NAc), dorsal hippocampus (DH) and medial prefrontal cortex (mPFC) may interact in memory processing for these two kinds of responses. Male Wistar
Mark C Tucci et al.
The European journal of neuroscience, 40(6), 2971-2979 (2014-06-18)
A previous analysis of the quinpirole sensitisation rat model of obsessive-compulsive disorder revealed that the behavioral phenotype of compulsive checking consists of three constitutive components - vigor of checking performance, focus on the task of checking, and satiety following a
Shaun Mehta et al.
JAMA facial plastic surgery, 16(6), 414-418 (2014-09-12)
This study evaluates the postoperative outcomes achieved with incisionless otoplasty for the correction of prominauris. To determine whether incisionless otoplasty is a reliable and replicable technique in correcting prominauris. This study consisted of a retrospective electronic medical record review for
S Picardi et al.
British journal of anaesthesia, 112(2), 370-379 (2013-09-26)
Signalling of several G-protein-coupled receptors of the Gq/11 family is time-dependently inhibited by local anaesthetics (LAs). Since LA-induced modulation of muscarinic m1 and m3 receptor function may explain their beneficial effects in clinical practice, such as decreased postoperative cognitive dysfunction

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service