Skip to Content
Merck
All Photos(3)

Documents

A1343

Sigma-Aldrich

Ammonium molybdate

meets USP testing specifications

Synonym(s):

Ammonium molybdate tetrahydrate, Ammonium heptamolybdate tetrahydrate, Molybdic acid ammonium salt tetrahydrate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(NH4)6Mo7O24 · 4H2O
CAS Number:
Molecular Weight:
1235.86
EC Number:
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:
NACRES:
NA.21

Agency

USP/NF
meets USP testing specifications

Quality Level

form

crystals

density

2.498 g/mL at 25 °C (lit.)

application(s)

pharmaceutical (small molecule)

SMILES string

N.N.N.N.N.N.O.O.O.O.O=[Mo](=O)=O.O=[Mo](=O)=O.O=[Mo](=O)=O.O=[Mo](=O)=O.O[Mo](O)(=O)=O.O[Mo](O)(=O)=O.O[Mo](O)(=O)=O

InChI

1S/7Mo.6H3N.10H2O.18O/h;;;;;;;6*1H3;10*1H2;;;;;;;;;;;;;;;;;;/q;;;;3*+2;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;/p-6

InChI key

FIXLYHHVMHXSCP-UHFFFAOYSA-H

Looking for similar products? Visit Product Comparison Guide

Application


  • Photochromic hydrogels for electronic applications: Ammonium molybdate was utilized in the synthesis of a polyacrylamide/sodium alginate/sodium chloride photochromic hydrogel. This hydrogel exhibits high conductivity, anti-freezing properties, and a fast response, making it suitable for information storage and electronic skin applications (Chen et al., 2024).

  • Microwave absorption materials: The preparation of flower-like carbon nanosheet aggregations embedded with ultrafine Mo(2)C, where ammonium molybdate plays a critical role, offers significant advancements in microwave absorption technology (Wang et al., 2023).

Storage Class Code

10-13 - German Storage Class 10 to 13

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Leandro Silva Costa et al.
Marine drugs, 9(6), 952-966 (2011-07-13)
Fucan is a term used to denominate a type of polysaccharide which contains substantial percentages of l-fucose and sulfate ester groups. We obtained five heterofucans from Sargassum filipendula by proteolytic digestion followed by sequential acetone precipitation. These heterofucans are composed
Sukyeong Lee et al.
Cell reports, 26(1), 29-36 (2019-01-04)
Hsp104 is a ring-forming, ATP-driven molecular machine that recovers functional protein from both stress-denatured and amyloid-forming aggregates. Although Hsp104 shares a common architecture with Clp/Hsp100 protein unfoldases, different and seemingly conflicting 3D structures have been reported. Examining the structure of
Orit Gat et al.
PLoS neglected tropical diseases, 5(11), e1373-e1373 (2011-11-10)
Antibiotic-resistant Salmonella enterica serovar Paratyphi A, the agent of paratyphoid A fever, poses an emerging public health dilemma in endemic areas of Asia and among travelers, as there is no licensed vaccine. Integral to our efforts to develop a S.
Pradeep Verma et al.
Bioresource technology, 102(4), 3941-3945 (2010-12-24)
Pretreatments for enzymatic saccharification are crucial for the establishment of lignocellulosic biorefineries. In this study, we focused on ammonium ions and peroxometal complexes as potential delignifying agents. We first examined the pretreatment of beech wood with nine different ammonium salts
Sidney Omelon et al.
Electrophoresis, 28(16), 2808-2811 (2007-07-21)
Nonradioactive polyphosphate (poly(P); (PO(3) (-))(n)) species resolved by PAGE can be detected by hydrolytic degradation of the polyphosphates into orthophosphates (P(i)) with a 5 M HCl solution saturated with NaCl, followed by staining the P(i) degradation products in a 1

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service