Merck
  • A nonradioactive method for detecting phosphates and polyphosphates separated by PAGE.
A nonradioactive method for detecting phosphates and polyphosphates separated by PAGE.
Electrophoresis (2007-07-21)
Sidney Omelon, Marc Grynpas
ABSTRACT
Nonradioactive polyphosphate (poly(P); (PO(3) (-))(n)) species resolved by PAGE can be detected by hydrolytic degradation of the polyphosphates into orthophosphates (P(i)) with a 5 M HCl solution saturated with NaCl, followed by staining the P(i) degradation products in a 1 M HCl solution of 0.25% w/v methyl green and 1% w/v ammonium molybdate. This method detects down to 0.5 nmol of phosphate as P(i), linear poly(P) (condensed phosphate), pyrophosphate (P(2)O(7) (4) (-)), or cyclic trimetaphosphate ion (P(3)O(9))(3) (-) species. This method improves the current method of staining linear poly(P) longer than four phosphate units with Toludine blue-O after PAGE. This study also shows that Stains-All can visualize resolved linear poly(P) shorter than those visualized by Toluidine blue-O. It is hoped that this sequential hydrolytic degradation and phosphate visualization method for detecting ortho-, linear, and cyclic poly(P) species will be a useful tool, as poly(P) are being discovered in a wide variety of biological systems, and their biochemical roles are still largely unknown.
Sigma-Aldrich
Ammonium molybdate tetrahydrate, BioUltra, ≥99.0% (T)
Pricing and availability is not currently available.
Sigma-Aldrich
Ammonium molybdate, 99.98% trace metals basis
Pricing and availability is not currently available.
Sigma-Aldrich
Ammonium molybdate tetrahydrate, BioReagent, suitable for cell culture, suitable for insect cell culture, 81.0-83.0% MoO3 basis
Pricing and availability is not currently available.
Sigma-Aldrich
Ammonium molybdate tetrahydrate, ACS reagent, 81.0-83.0% MoO3 basis
Pricing and availability is not currently available.
Sigma-Aldrich
Ammonium molybdate tetrahydrate, for inorganic trace analysis, ACS reagent, 99.98% trace metals basis
Pricing and availability is not currently available.
Sigma-Aldrich
Ammonium molybdate, meets USP testing specifications
Pricing and availability is not currently available.