Skip to Content
Merck
All Photos(3)

Key Documents

221759

Sigma-Aldrich

Ammonium cerium(IV) sulfate dihydrate

Synonym(s):

Ceric ammonium sulfate, Cerium(IV) ammonium sulfate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Ce(NH4)4(SO4)4 · 2H2O
CAS Number:
Molecular Weight:
632.55
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23
grade:
for analytical purposes
form:
powder
Pricing and availability is not currently available.

grade

for analytical purposes

Assay

≥94.0% (with KMnO4, redox titration)

form

powder

composition

Ce(IV), ≥20%

reaction suitability

reagent type: catalyst
core: cerium

mp

130 °C (lit.)

SMILES string

N.N.N.N.O.O.[Ce+4].OS([O-])(=O)=O.OS([O-])(=O)=O.OS([O-])(=O)=O.OS([O-])(=O)=O

InChI

1S/Ce.4H3N.4H2O4S.2H2O/c;;;;;4*1-5(2,3)4;;/h;4*1H3;4*(H2,1,2,3,4);2*1H2/q+4;;;;;;;;;;/p-4

InChI key

VCNAMBGKEDPVGQ-UHFFFAOYSA-J

Looking for similar products? Visit Product Comparison Guide

Application

The product may be used to initiate the radical polymerization of pectin-poly(ethylene glycol) methacrylate (PEGMA) supracmolecular hydrogels.[1] Ammonium cerium sulphate impregnated starch may be carbonized to form cerium dispersed carbon (CeDC) sorbent used for the extraction of excess fluoride in drinking water.[2]

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Thixotropic Supramolecular Pectin-Poly (Ethylene Glycol) Methacrylate (PEGMA) Hydrogels.
Chan SY, et al.
Polymer, 8(11), 404-404 (2016)
Cerium dispersed in carbon (CeDC) and its adsorption behavior: A first example of tailored adsorbent for fluoride removal from drinking water
Sivasankar V, et al.
Chemical Engineering Journal, 214, 45-54 (2013)
S Kaul et al.
Amino acids, 34(2), 315-320 (2006-11-07)
An assessment of the potential of proline to scavenge free radicals was made in a couple of in vitro assay systems, namely graft co-polymerization and autooxidation of pyrogallol. Both these assays are essentially dependent upon free radical mechanisms. Graft co-polymerization
K H Gordon et al.
Organic letters, 3(1), 53-56 (2001-06-30)
[figure: see text] A novel benzyloxyaniline linker that uses ceric ammonium nitrate (CAN) as a cleavage reagent is described. Its application in the solid-phase synthesis of N-unsubstituted beta-lactams and secondary amides furnishes compounds in moderate to excellent yield (45-91%) and
C Passirani et al.
Journal of biomaterials science. Polymer edition, 10(1), 47-62 (1999-03-26)
Nanoparticles have been obtained directly in aqueous media, from amphiphilic copolymers synthesized by radical polymerization of methyl methacrylate (MMA) initiated by Ce(IV) ions in the presence of heparin or dextran. The reaction conditions under which the copolymers spontaneously formed nanoparticles

Questions

1–2 of 2 Questions  
  1. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

  2. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service