Skip to Content
Merck
All Photos(2)

Documents

93878

Supelco

trans-Sinapic acid

analytical standard

Synonym(s):

trans-4-Hydroxy-3,5-dimethoxy-cinnamic acid, trans-Sinapinic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H12O5
CAS Number:
Molecular Weight:
224.21
EC Number:
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

Assay

≥99.0% (HPLC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

mp

195-205 °C (dec.)

application(s)

food and beverages

format

neat

SMILES string

COc1cc(\C=C\C(O)=O)cc(OC)c1O

InChI

1S/C11H12O5/c1-15-8-5-7(3-4-10(12)13)6-9(16-2)11(8)14/h3-6,14H,1-2H3,(H,12,13)/b4-3+

InChI key

PCMORTLOPMLEFB-ONEGZZNKSA-N

Looking for similar products? Visit Product Comparison Guide

Application

trans-Sinapic acid may be used as an analytical reference standard for the quantification of the analyte in seed samples and vegetables using different chromatography techniques.
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Other Notes

This compound is commonly found in plants of the genus: carum cinnamomum melissa salvia thymus zingiber

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Quantitation of free and hydrolyzable phenolic acids in seeds by capillary gas-liquid chromatography
Dabrowski.JK and Sosulski.WF
Journal of Agricultural and Food Chemistry, 32(1), 123-127 (1984)
Free and soluble bound phenolic acids in some cruciferous vegetables
Uda Y, et al.
Journal of the Japanese Society for Food Science and Technology, 35(5), 360-366 (1988)
Jacob C Dean et al.
Journal of the American Chemical Society, 136(42), 14780-14795 (2014-10-09)
Ultraviolet spectroscopy of sinapoyl malate, an essential UV-B screening agent in plants, was carried out in the cold, isolated environment of a supersonic expansion to explore its intrinsic UV spectral properties in detail. Despite these conditions, sinapoyl malate displays anomalous
Jiyan Liu et al.
Journal of ethnopharmacology, 155(2), 1322-1331 (2014-07-30)
Chemotherapy is the mainstay of modern tuberculosis (TB) control. Traditional Chinese Medicine (TCM) can enhance the effect of anti-TB drug, promote the absorption of the foci in the lung and reduce drug toxicity. In TCM, the determination of treatment is
Stefany Grutzmann Arcari et al.
Electrophoresis, 41(20), 1784-1792 (2020-08-12)
This study aimed to evaluate the polyphenolic composition along with the biological activity of guabiroba (Campomanesia xanthocarpa Berg.) fruits using comprehensive two-dimensional liquid chromatography (LC × LC). A simplex centroid design comprising three solvents (methanol, 2% acetic acid, and acetonitrile) was used

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service