Skip to Content
Merck
All Photos(1)

Key Documents

17793

Sigma-Aldrich

Quercetin 3-β-D-glucoside

≥90% (HPLC)

Synonym(s):

3,3′,4′,5,7-Pentahydroxyflavone 3-β-glucoside, Isoquercitrin

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H20O12
CAS Number:
Molecular Weight:
464.38
Beilstein:
100989
MDL number:
UNSPSC Code:
12352205
PubChem Substance ID:
NACRES:
NA.25

Assay

≥90% (HPLC)

application(s)

metabolomics
vitamins, nutraceuticals, and natural products

storage temp.

−20°C

SMILES string

OC[C@H]1O[C@@H](OC2=C(Oc3cc(O)cc(O)c3C2=O)c4ccc(O)c(O)c4)[C@H](O)[C@@H](O)[C@@H]1O

InChI

1S/C21H20O12/c22-6-13-15(27)17(29)18(30)21(32-13)33-20-16(28)14-11(26)4-8(23)5-12(14)31-19(20)7-1-2-9(24)10(25)3-7/h1-5,13,15,17-18,21-27,29-30H,6H2/t13-,15-,17+,18-,21+/m1/s1

InChI key

OVSQVDMCBVZWGM-QSOFNFLRSA-N

Gene Information

mouse ... Hexa(15211)

Looking for similar products? Visit Product Comparison Guide

General description

Quercetin 3-β-D-glucoside (Q3G), also known as isoquercitrin, is one of the major glycosidic forms of quercetin. It is a mono-glucoside, first isolated from the seed pods of Cercis canadensis L., it is also found in various plants, vegetables, and fruits. Q3G has the basic chemical structure of a flavonoid, with a glucose molecule attached to the C-3 of quercetin.

Application

Quercetin 3-β-D-glucoside has been used:
  • as a dietary flavonoid supplement to check its binding capacity with human small ubiquitin-related modifier 1 (SUMO1) protein using surface plasmon resonance (SPR)
  • as an inhibitor for Escherichia coli adenosine triphosphate (ATP) synthase
  • as an anti-aggregation agent to test its activity against β-amyloid, green fluorescent protein (GFP), and chymotrypsinogen proteins

Biochem/physiol Actions

Quercetin 3-β-D-glucoside possesses strong antioxidant and anti-inflammatory activities. Being a potent oxygen-radical scavenger, it is a neuroprotective agent that improves Alzheimer′s disease (AD) condition in mice models. It has shown antiproliferative activity against colon, lung, and hepatocellular cell lines and MCF-7 human breast cancer cells in combination with apple extracts.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Prasanna K Dadi et al.
International journal of biological macromolecules, 45(1), 72-79 (2009-04-21)
We have studied the inhibitory effect of five polyphenols namely, resveratrol, piceatannol, quercetin, quercetrin, and quercetin-3-beta-D glucoside on Escherichia coli ATP synthase. Recently published X-ray crystal structures of bovine mitochondrial ATP synthase inhibited by resveratrol, piceatannol, and quercetin, suggest that
Jun Yang et al.
Journal of agricultural and food chemistry, 57(18), 8581-8586 (2009-08-22)
Breast cancer is the most frequently diagnosed cancer in women. An alternative strategy to reduce the risk of cancer is through dietary modification. Although phytochemicals naturally occur as complex mixtures, little information is available regarding possible additive, synergistic, or antagonistic
Kateřina Valentová et al.
Food and chemical toxicology : an international journal published for the British Industrial Biological Research Association, 68, 267-282 (2014-04-01)
The flavonoid isoquercitrin (quercetin-3-O-β-d-glucopyranoside) is commonly found in medicinal herbs, fruits, vegetables and plant-derived foods and beverages. This article reviews the occurrence, preparation, bioavailability, pharmacokinetics, toxicology and biological activity of isoquercitrin and "enzymatically modified (α-glucosylated) isoquercitrin" (EMIQ). Pure isoquercitrin can
Quercetin and quercetin-3-beta-d-glucoside improve cognitive and memory function in Alzheimer?s disease mouse
Kim J H, et al.,
Applied Biological Chemistry, 721-728 (2016)
Hyun Ju You et al.
Journal of agricultural and food chemistry, 58(20), 10886-10892 (2010-10-05)
The flavonol quercetin in plants and foods occurs predominantly in the form of glycoside whose sugar moiety affects the bioavailability and the mechanism of its biological activities. The antiproliferative activities of quercetin derivatives such as quercetin aglycone, quercetin-3-β-D-glucoside (Q3G), and

Protocols

HPLC Analysis of Polyphenols in Nero d'Avola Red Wine on Discovery® HS C18 (UV 280 nm)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service