Skip to Content
Merck
All Photos(3)

Key Documents

R4033

Sigma-Aldrich

Rosmarinic acid

≥98% (HPLC), from Rosemarinus officinalis L.

Synonym(s):

(R)-O-(3,4-Dihydroxycinnamoyl)-3-(3,4- dihydroxyphenyl)lactic acid, 3,4-Dihydroxycinnamic acid (R)-1-carboxy-2-(3,4-dihydroxyphenyl)ethyl ester

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H16O8
CAS Number:
Molecular Weight:
360.31
Beilstein:
2227587
MDL number:
UNSPSC Code:
12352106
PubChem Substance ID:
NACRES:
NA.25

biological source

Rosemarinus officinalis L.

Quality Level

Assay

≥98% (HPLC)

form

powder

storage condition

protect from light

color

white to faint beige

mp

171-175 °C (lit.)

solubility

H2O: 1 mg/mL, clear, colorless

application(s)

metabolomics
vitamins, nutraceuticals, and natural products

storage temp.

2-8°C

SMILES string

OC(=O)[C@@H](Cc1ccc(O)c(O)c1)OC(=O)\C=C\c2ccc(O)c(O)c2

InChI

1S/C18H16O8/c19-12-4-1-10(7-14(12)21)3-6-17(23)26-16(18(24)25)9-11-2-5-13(20)15(22)8-11/h1-8,16,19-22H,9H2,(H,24,25)/b6-3+/t16-/m1/s1

InChI key

DOUMFZQKYFQNTF-WUTVXBCWSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Rosmarinic acid is a naturally occurring polyphenolic compound found in plants of the Boraginaceae family and Nepetoideae subfamily. It is an ester of caffeic acid as well as 3,4-dihydroxy-phenyllactic acid.

Application

Rosmarinic acid has been used:
  • to see its protective effects on the human endothelial cell line (EA.hy926) to treat diabetic atherosclerosis
  • to test its effect in 1-Methyl-4-phenyl-1, 2, 3, 6-tetrahydropyridine (MPTP) induced Parkinson′s disease (PD) models on attenuating inflammatory responses and microglia activation
  • to test its neuroprotective effects on dopaminergic neurons
  • to test its effects on sperm motility, viability and on intracellular calcium ion concentration
  • as a reference standard in capillary zone electrophoresis (CZE) method

Biochem/physiol Actions

Rosmarinic acid (RosA) is a well-appreciated agent used in several diabetic conditions and complications such as diabetic nephropathy, diabetic neuropathy and diabetic atherosclerosis. It has also been implicated to be a neuroprotective agent for several neurological disorders such as Alzheimer′s disease, Parkinson′s disease, and epilepsy by suppressing their conditions.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Meng-Ge Lv et al.
Reproductive toxicology (Elmsford, N.Y.), 81, 58-63 (2018-07-17)
Rosmarinic acid (RA), a natural phenolic ester, is cytoprotective for male reproduction in animal models. The present study investigated the in vitro actions of RA on human sperm functions. Human sperm were exposed to 1, 10, 100, and 1000 μM
Polyphenols analysis from different medicinal plants extracts using capillary zone electrophoresis (CZE)
Matei AO, et al.,
Revista de Chimie, 67, 1051-1105 (2016)
Tomoki Hase et al.
Scientific reports, 9(1), 8711-8711 (2019-06-20)
A new mechanism is revealed by which a polyphenol, rosmarinic acid (RA), suppresses amyloid β (Aβ) accumulation in mice. Here we examined the brains of mice (Alzheimer's disease model) using DNA microarray analysis and revealed that the dopamine (DA)-signaling pathway
A Oğuz et al.
Folia morphologica, 79(2), 288-295 (2019-08-23)
The aim of the study was to investigate the protective effects of rosmarinic acid in rats exposed to hepatic ischaemia/reperfusion (I/R) injury. Thirty-two rats were randomly classified into four groups of 8 rats each: laparotomy without medication, rosmarinic acid (dose
Eun-Ju Yang et al.
Journal of cellular physiology, 228(5), 975-982 (2012-10-09)
High mobility group box 1 (HMGB1) protein is a crucial cytokine that mediates response to infection, injury, and inflammation. Rosmarinic acid (RA) is an important component of the leaves of Perilla frutescens and has neuroprotective, anti-microbial, anti-oxidant, and anti-cancer effects

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service