Skip to Content
Merck
All Photos(1)

Key Documents

90820

Sigma-Aldrich

Tributyl phosphate

puriss., ≥99.0% (GC)

Synonym(s):

TBP, TBPA

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3(CH2)3O)3PO
CAS Number:
Molecular Weight:
266.31
Beilstein:
1710584
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

vapor density

9.2 (vs air)

Quality Level

vapor pressure

27 mmHg ( 178 °C)
7.3 mmHg ( 150 °C)

grade

puriss.

Assay

≥99.0% (GC)

form

liquid

autoignition temp.

770 °F

refractive index

n20/D 1.424 (lit.)

bp

180-183 °C/22 mmHg (lit.)

mp

−79 °C (lit.)

density

0.979 g/mL at 25 °C (lit.)

functional group

phosphate

SMILES string

CCCCOP(=O)(OCCCC)OCCCC

InChI

1S/C12H27O4P/c1-4-7-10-14-17(13,15-11-8-5-2)16-12-9-6-3/h4-12H2,1-3H3

InChI key

STCOOQWBFONSKY-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Tributyl phosphate (TBP) is an organophosphorus compound. TBP is a strong Lewis base and interacts with acids. It is used as a solvent for rare earth extraction and the nuclear fuel cycle. Additionally, TBP is used as a plasticizer for cellulose esters, lacquers, plastics, and vinyl resins

Application

Tributyl phosphate is used as:
  • An extractant for monocarboxylic acids(formic, acetic, and propionic) from an aqueous medium.
  • An additive to enhance the performance of dye-sensitized solar cells (DSSCs).
  • A solvent in combination with ionic liquid[1-butyl-3-methyl imidazolium tetrafluoroborate] for the extraction of phenol and chlorophenols.

Pictograms

Health hazardExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 3 - Carc. 2 - Skin Irrit. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

293.0 °F - closed cup

Flash Point(C)

145 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Detailed investigation of lactic acid extraction with tributylphosphate dissolved in dodecane
Labbaci A, et al.
Journal of Chemical and Engineering Data, 55(1), 228-233 (2010)
Extraction of phenol and chlorophenols using ionic liquid [Bmim]+[BF4]- dissolved in tributyl phosphate
Brinda Lakshmi A, et al.
Clean: Soil, Air, Water, 41(4), 349-355 (2013)
Enhanced photovoltaic properties of modified redox electrolyte in dye-sensitized solar cells using tributyl phosphate as additive
Afrooz M and Dehghani H
Journal of Power Sources, 262, 140-146 (2014)
Reactive extraction of monocarboxylic acids (formic, acetic, and propionic) using tributyl phosphate in green solvents (cyclopentyl methyl ether and 2-methyltetrahydrofuran
Tu?rk FN, et al.
Journal of Chemical and Engineering Data, 66(1), 130-137 (2020)
Omar Arous et al.
Journal of hazardous materials, 180(1-3), 493-498 (2010-05-19)
In this work, a development of polymeric inclusion membranes for the cations separation is reported. The membrane was made up of cellulose triacetate (CTA) with a tributyl phosphate (TBP) incorporated into the polymer as metal ions carrier. The transport of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service