Skip to Content
Merck
All Photos(2)

Key Documents

240494

Sigma-Aldrich

Tributyl phosphate

≥99%

Synonym(s):

TBP, TBPA

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3(CH2)3O)3PO
CAS Number:
Molecular Weight:
266.31
Beilstein:
1710584
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Pricing and availability is not currently available.

vapor density

9.2 (vs air)

Quality Level

vapor pressure

27 mmHg ( 178 °C)
7.3 mmHg ( 150 °C)

Assay

≥99%

form

liquid

autoignition temp.

770 °F

refractive index

n20/D 1.424 (lit.)

bp

180-183 °C/22 mmHg (lit.)

mp

−79 °C (lit.)

solubility

organic solvents: miscible
water: soluble (1mL in 165mL)

density

0.979 g/mL at 25 °C (lit.)

functional group

phosphate

SMILES string

CCCCOP(=O)(OCCCC)OCCCC

InChI

1S/C12H27O4P/c1-4-7-10-14-17(13,15-11-8-5-2)16-12-9-6-3/h4-12H2,1-3H3

InChI key

STCOOQWBFONSKY-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

The microbial degradation of tributyl phosphate was carried out using Klebsiella pneumoniae S3 isolated from the soil[1]. The solubilization behavior of TBP in aqueous solutions of L64-Pluronics was studied using light and small angle neutron scattering (SANS)[2].

Tributyl phosphate (TBP) is an organophosphate compound widely used as an extraction solvent, flame retardant, and plasticizer. [3]

Application

Tributyl phosphate (TBP) was used as defoamers and/or plasticizers, in fuel reprocessing and extraction[1].

Pictograms

Health hazardExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 3 - Carc. 2 - Skin Irrit. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

293.0 °F - closed cup

Flash Point(C)

145 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Norela Jusoh et al.
Environmental science and pollution research international, 27(18), 23246-23257 (2020-04-27)
Polluted sterilization condensate discharged from palm oil mill may contain polyphenols that are rich in the antioxidant property. Emulsion liquid membrane (ELM) process is a promising method for polyphenol recovery due to its several attractive features such as high selectivity
J Causse et al.
Langmuir : the ACS journal of surfaces and colloids, 26(20), 15745-15753 (2010-09-30)
We have studied the solubilization behavior of tributylphosphate (TBP) in aqueous solutions of L64-Pluronics, using light and small angle neutron scattering (SANS). Varying the temperature and the oil content, the system presents a nontrivial phase behavior. In particular, at 308
Lubrication on the curve surface structure using palm oil and mineral oil
Razak, D Md and Syahrullail, et al.
Procedia Engineering, 68, 607-612 (2013)
Padryk Merkl et al.
Biosensors & bioelectronics, 171, 112732-112732 (2020-10-30)
Biofilms are dense bacterial colonies that may adhere to the surfaces of medical devices and are major contributors to infections. These colonies are characterized by a self-produced matrix of extracellular polymeric substances (EPS). Bacterial biofilms are difficult to treat with
S V Kulkarni et al.
Applied microbiology and biotechnology, 98(2), 919-929 (2013-05-07)
Tributyl phosphate (TBP) has enormous applications in the field of extraction, fuel reprocessing, as defoamers and/or plasticizers. Excessive usage of this organophosphorus compound, poses an environmental threat. The present study deals with microbial degradation of TBP using Klebsiella pneumoniae S3

Questions

  1. Can you make any comment on the stability of TBP when diluted in phosphate-buffered saline (1% v/v)? Would it be recommended to store such a diluted solution in the fridge?

    1 answer
    1. The stability of the product in solution has not been determined. Most publications reference TBP solutions in organic solvents rather than aqueous solutions. The following paper, while very dated, may offer some additional information: https://www.osti.gov/servlets/purl/4349304

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service