Skip to Content
Merck
All Photos(1)

Key Documents

00675

Supelco

Tributyl phosphate

for extraction analysis, ≥99.0% (GC)

Synonym(s):

TBP, TBPA

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3(CH2)3O)3PO
CAS Number:
Molecular Weight:
266.31
Beilstein:
1710584
EC Number:
MDL number:
UNSPSC Code:
41116105
PubChem Substance ID:
NACRES:
NA.21
technique(s):
GC/GC: suitable
bp:
180-183 °C/22 mmHg (lit.)
vapor pressure:
27 mmHg ( 178 °C)
7.3 mmHg ( 150 °C)
Pricing and availability is not currently available.

vapor density

9.2 (vs air)

Quality Level

vapor pressure

27 mmHg ( 178 °C)
7.3 mmHg ( 150 °C)

Assay

≥99.0% (GC)

form

liquid

autoignition temp.

770 °F

technique(s)

GC/GC: suitable

refractive index

n20/D 1.424 (lit.)
n20/D 1.424

bp

180-183 °C/22 mmHg (lit.)

mp

−79 °C (lit.)

acid value

<1

density

0.979 g/mL at 25 °C (lit.)

cation traces

Al: ≤10 mg/kg
Ba: ≤1 mg/kg
Bi: ≤1 mg/kg
Ca: ≤5 mg/kg
Cd: ≤0.5 mg/kg
Co: ≤0.5 mg/kg
Cr: ≤1 mg/kg
Cu: ≤0.5 mg/kg
Fe: ≤5 mg/kg
K: ≤5 mg/kg
Li: ≤1 mg/kg
Mg: ≤1 mg/kg
Mn: ≤0.5 mg/kg
Mo: ≤1 mg/kg
Na: ≤5 mg/kg
Ni: ≤0.5 mg/kg
Pb: ≤1 mg/kg
Sr: ≤1 mg/kg
Zn: ≤1 mg/kg

SMILES string

CCCCOP(=O)(OCCCC)OCCCC

InChI

1S/C12H27O4P/c1-4-7-10-14-17(13,15-11-8-5-2)16-12-9-6-3/h4-12H2,1-3H3

InChI key

STCOOQWBFONSKY-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Health hazardExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 3 - Carc. 2 - Skin Irrit. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

293.0 °F - closed cup

Flash Point(C)

145 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

T Burnouf et al.
Vox sanguinis, 104(1), 1-6 (2012-07-05)
A minipool solvent/detergent (S/D; 1% TnBP/1% Triton X-45; 31°C) process was developed for viral inactivation of plasma and cryoprecipitate used for transfusion. The goal of this study was to determine the rate and extent of inactivation of dengue virus (DENV)
Domenico Santoro et al.
Environmental science & technology, 44(16), 6233-6241 (2010-08-14)
Advanced Oxidation Processes (AOPs) promoted by ultraviolet light are innovative and potentially cost-effective solutions for treating persistent pollutants recalcitrant to conventional water and wastewater treatment. While several studies have been performed during the past decade to improve the fundamental understanding
C W Van Neste et al.
Analytical chemistry, 81(5), 1952-1956 (2009-02-04)
Despite its immediate applications, selective detection of trace quantities of surface adsorbed chemicals, such as explosives, without physically collecting the sample molecules is a challenging task. Standoff spectroscopic techniques offer an ideal method of detecting chemicals without using a sample
Muniasamy Neerathilingam et al.
Toxicology letters, 199(1), 10-16 (2010-08-07)
Tributyl phosphate (TBP) is a toxic organophosphorous compound widely used in many industrial applications, including significant usage in nuclear processing. The industrial application of this chemical is responsible for occupational exposure and environmental pollution. In this study, (1)H NMR-based metabonomics
Torsten Klengel et al.
Nature neuroscience, 16(1), 33-41 (2012-12-04)
Although the fact that genetic predisposition and environmental exposures interact to shape development and function of the human brain and, ultimately, the risk of psychiatric disorders has drawn wide interest, the corresponding molecular mechanisms have not yet been elucidated. We

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service