Skip to Content
Merck
All Photos(1)

Documents

63081

Sigma-Aldrich

Magnesium hydroxide

BioUltra, ≥99.0% (KT)

Synonym(s):

Magnesium dihydroxide, Mg(OH)2

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Mg(OH)2
CAS Number:
Molecular Weight:
58.32
EC Number:
MDL number:
UNSPSC Code:
12352303
PubChem Substance ID:
NACRES:
NA.25

product line

BioUltra

Quality Level

Assay

≥99.0% (KT)

impurities

Insoluble matter, passes filter test

mp

350 °C (lit.)

solubility

5 M HCl: 0.1 M at 20 °C, clear, colorless

anion traces

chloride (Cl-): ≤500 mg/kg
sulfate (SO42-): ≤1000 mg/kg

cation traces

As: ≤0.1 mg/kg
Ba: ≤10 mg/kg
Bi: ≤50 mg/kg
Ca: ≤5000 mg/kg
Cd: ≤5 mg/kg
Co: ≤5 mg/kg
Cr: ≤5 mg/kg
Cu: ≤5 mg/kg
Fe: ≤100 mg/kg
K: ≤50 mg/kg
Li: ≤5 mg/kg
Mn: ≤10 mg/kg
Mo: ≤5 mg/kg
Na: ≤100 mg/kg
Ni: ≤100 mg/kg
Pb: ≤5 mg/kg
Sr: ≤10 mg/kg
Zn: ≤5 mg/kg

λ

0.1 M in 5 M HCl

UV absorption

λ: 260 nm Amax: 0.030
λ: 280 nm Amax: 0.025

SMILES string

O[Mg]O

InChI

1S/Mg.2H2O/h;2*1H2/q+2;;/p-2

InChI key

VTHJTEIRLNZDEV-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

General description

Magnesium hydroxide is commonly used as a non-systemic antacid for treating acid-peptic disorders. It helps in the treatment kidney stones patients by reducing the recurrence rate.

Application

Magnesium hydroxide is used as a dietary supplement to study its effects on the absorbtion of factors such as folic acid and iron and the release of the peptide hormone cholecystokinin (CCK).

Storage Class Code

13 - Non Combustible Solids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

G Johansson et al.
The Journal of urology, 124(6), 770-774 (1980-12-01)
Prophylactic treatment with magnesium hydroxide ws instituted in 56 consecutive cases with renal calcium stones. The patients had been investigated previously with regard to the magnesium metabolism. The urinary magnesium excretion increased promptly and remained on a higher level during
R M Russell et al.
The Journal of laboratory and clinical medicine, 112(4), 458-463 (1988-10-01)
Intestinal folic acid transport is a saturable process with a pH optimum of 5.5 to 6.0. Because of the possible effects of antacids and acid-lowering drugs on the pH of the proximal small intestine, the influence of these drugs on
B K Snyder et al.
Annals of emergency medicine, 33(4), 400-405 (1999-03-27)
Animal models have demonstrated that administration of magnesium hydroxide (MgOH) decreases serum iron levels after an iron overdose. We designed this study to determine the effect of MgOH administration on iron absorption and adverse effects after a supratherapeutic dose of
Tengfei Lv et al.
Journal of hazardous materials, 237-238, 121-132 (2012-09-11)
The present study explores the potential of Mg-Ca-Al (NO(3)) hydrotalcite-like compounds (MgCaAlNO(3)-HTlcs) for the removal of fluoride from protein solutions. In this study, the Mg(3-x)Ca(x)AlNO(3)-HTlcs (x=0-3, x is the mol.% of Ca) were synthesized and characterized by SEM, XRD, FTIR
Anna L Harrison et al.
Environmental science & technology, 47(1), 126-134 (2012-07-10)
Atmospheric CO(2) is sequestered within ultramafic mine tailings via carbonation of Mg-bearing minerals. The rate of carbon sequestration at some mine sites appears to be limited by the rate of CO(2) supply. If carbonation of bulk tailings were accelerated, large

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service