M7179
Magnesium carbonate
meets USP testing specifications
Synonym(s):
Magnesium carbonate hydrate
Sign Into View Organizational & Contract Pricing
All Photos(2)
About This Item
Linear Formula:
MgCO3 · x H2O
CAS Number:
Molecular Weight:
84.31 (anhydrous basis)
MDL number:
UNSPSC Code:
51171508
PubChem Substance ID:
NACRES:
NA.21
Recommended Products
Agency
USP/NF
meets USP testing specifications
Quality Level
concentration
40.0-43.5% (Mg (as MgO))
application(s)
pharmaceutical (small molecule)
SMILES string
O.[Mg++].[O-]C([O-])=O
InChI
1S/CH2O3.Mg.H2O/c2-1(3)4;;/h(H2,2,3,4);;1H2/q;+2;/p-2
InChI key
OUHCLAKJJGMPSW-UHFFFAOYSA-L
Looking for similar products? Visit Product Comparison Guide
Application
- Growth, secondary metabolite production, and in vitro antiplasmodial activity of Sonchus arvensis L. callus under dolomite [CaMg(CO3)2] treatment.: This research investigates the effect of dolomite, containing magnesium carbonate, on the growth and biochemical profile of Sonchus arvensis L. callus cultures, highlighting its potential for enhancing bioactive compound production (Wahyuni et al., 2021).
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Nadine Kaesler et al.
Nephrology, dialysis, transplantation : official publication of the European Dialysis and Transplant Association - European Renal Association, 35(1), 65-73 (2019-02-05)
Optimal phosphate control is an unmet need in chronic kidney disease (CKD). High serum phosphate increases calcification burden and is associated with mortality and cardiovascular disease in CKD. Nicotinamide (NA) alone or in combination with calcium-free phosphate binders might be
Elijah J Petersen et al.
Environmental health perspectives, 116(4), 496-500 (2008-04-17)
Carbon nanotubes represent a class of nanomaterials having broad application potentials and documented cellular uptake and ecotoxicological effects that raise the possibility that they may bioaccumulate in living organisms. Radioactively labeled nanotubes were synthesized using a novel methane chemical vapor
Ronak B Shah et al.
Journal of controlled release : official journal of the Controlled Release Society, 196, 60-70 (2014-09-16)
A biomimetic approach to organic solvent-free microencapsulation of proteins based on the self-healing capacity of poly (DL)-lactic-co-glycolic acid (PLGA) microspheres containing glycosaminoglycan-like biopolymers (BPs), was examined. To screen BPs, aqueous solutions of BP [high molecular weight dextran sulfate (HDS), low
Johanna Mosig et al.
Journal of pharmaceutical sciences, 104(3), 1108-1118 (2015-01-07)
The influence of lubrication and particle size on the reduced compactability after dry granulation was investigated. Powder cellulose, lactose, magnesium carbonate, and two types of microcrystalline cellulose were roll compacted, granulated, and sieved into particle fractions. Particle fractions were compressed
Benjamin T Manard et al.
Applied spectroscopy, 69(1), 58-66 (2014-12-17)
Liquid sampling-atmospheric pressure glow discharge (LS-APGD) microplasma is being developed as a secondary vaporization-excitation source for the optical emission analysis of laser ablation (LA)-generated particle populations. The practicalities of this coupling are evaluated by determining the influence of source parameters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service