Skip to Content
Merck
All Photos(1)

Key Documents

63069

Sigma-Aldrich

Magnesium chloride solution

Magnesium chloride solution
1 of 1 reviewers received a sample product or took part in a promotion

BioUltra, for molecular biology, ~1 M in H2O

Synonym(s):

Magnesium Chloride hydrate, Magnesium chloride (MgCl2)hexahydrate, MgCl2

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
MgCl2
CAS Number:
Molecular Weight:
95.21
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.26
grade:
Molecular Biology
for molecular biology
form:
liquid
solubility:
water: soluble
Pricing and availability is not currently available.

grade

Molecular Biology
for molecular biology

Quality Level

product line

BioUltra

form

liquid

concentration

~1 M in H2O

technique(s)

HPLC: suitable
flow cytometry: suitable

impurities

DNases, none detected
RNases, none detected
insoluble matter, passes filter test
phosphatases, none detected
proteases, none detected
≤0.0002% total Nitrogen (N)

pH

5.0-7.5 (25 °C, 1 M in H2O)

solubility

water: soluble

density

1.08 g/mL at 20 °C

anion traces

phosphate (PO43-): ≤10 mg/kg
sulfate (SO42-): ≤50 mg/kg

cation traces

Al: ≤1 mg/kg
As: ≤0.1 mg/kg
Ba: ≤1 mg/kg
Bi: ≤1 mg/kg
Ca: ≤10 mg/kg
Cd: ≤1 mg/kg
Co: ≤1 mg/kg
Cr: ≤1 mg/kg
Cu: ≤1 mg/kg
Fe: ≤1 mg/kg
K: ≤50 mg/kg
Li: ≤1 mg/kg
Mn: ≤1 mg/kg
Mo: ≤1 mg/kg
Na: ≤500 mg/kg
Ni: ≤1 mg/kg
Pb: ≤1 mg/kg
Sr: ≤1 mg/kg
Zn: ≤1 mg/kg

λ

1 M in H2O

UV absorption

λ: 260 nm Amax: 0.05
λ: 280 nm Amax: 0.03

suitability

suitable for molecular biology

SMILES string

Cl[Mg]Cl

application(s)

cell analysis
filtration
vaccine development

InChI

1S/2ClH.Mg/h2*1H;/q;;+2/p-2

InChI key

TWRXJAOTZQYOKJ-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

General description

Magnesium chloride is widely used in polymerase chain reactions (PCR) buffers to supply magnesium ions. The concentration of Mg2+ is the most critical parameter that impacts both the efficiency and specificity of PCR when total genomic DNA is used, synthesis of long products, or during multiplex reactions. Elevated concentrations of Mg2+ enhance the stability of primer-template duplexes and the efficiency of amplification.[1]

Application

Magnesium chloride solution has been used as a component of nuclei isolation buffer for in situ fixations and isolation of nuclei from Kupffer cells (or other liver cells).[2]
Magnesium chloride solution has been used in the reaction mix of reverse transcriptase enzyme, PFV RT.[3] It has been used in the preparation of artificial cerebrospinal fluid (ACSF).[4]

Other Notes

For additional information on our range of Biochemicals, please complete this form.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

RNase H Polymerase-independent Cleavage Assay for Evaluation of RNase H Activity of Reverse Transcriptase Enzymes.
Corona A and Tramontano E
Bio-protocol null
Benjamin Judkewitz et al.
Nature protocols, 4(6), 862-869 (2009-05-16)
In order to link our knowledge of single neurons with theories of network function, it has been a long-standing goal to manipulate the activity and gene expression of identified subsets of mammalian neurons within the intact brain in vivo. This
Ty D Troutman et al.
STAR protocols, 2(1), 100363-100363 (2021-03-23)
Significant advancements in understanding disease mechanisms can occur through combined analysis of next-generation sequencing datasets generated using purified cell populations. Here, we detail our optimized protocol for purification of mouse hepatic macrophages (or other liver non-parenchymal populations) suitable for use
Elaine T Lim et al.
PLoS genetics, 10(7), e1004494-e1004494 (2014-08-01)
Exome sequencing studies in complex diseases are challenged by the allelic heterogeneity, large number and modest effect sizes of associated variants on disease risk and the presence of large numbers of neutral variants, even in phenotypically relevant genes. Isolated populations
Hideyuki Arita et al.
Brain tumor pathology, 32(1), 22-30 (2014-04-22)
Assessment of the mutational status of the isocitrate dehydrogenase 1/2 (IDH1/2) gene has become an integral part of the standard diagnostic procedure and, therefore, needs to be accurate. This may, however, be compromised by various factors including the method of

Questions

1–2 of 2 Questions  
  1. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

  2. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

Reviews

1 of 1 reviewers received a sample product or took part in a promotion

Active Filters

  1. Switzerland
    • Reviews 2
    • Votes 0
    5 out of 5 stars.

    Good as ever

    It is now in a plastic bottle, not in the glass anymore but it still works nicely

    Helpful?

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service