Skip to Content
Merck
All Photos(1)

Key Documents

8.14733

Sigma-Aldrich

Magnesium chloride

anhydrous for synthesis

Synonym(s):

Magnesium chloride, Magnesium dichloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
Cl2Mg
CAS Number:
Molecular Weight:
95.21
MDL number:
UNSPSC Code:
12352302
EC Index Number:
232-094-6
NACRES:
NA.22
grade:
for synthesis
synthesis grade
form:
solid
solubility:
542 g/L
Pricing and availability is not currently available.

grade

for synthesis
synthesis grade

Quality Level

form

solid

potency

2800 mg/kg LD50, oral (Rat)

pH

≥7 (20 °C, 50 g/L in H2O)

bp

1412 °C/1013 hPa

mp

712 °C

solubility

542 g/L

density

2.32 g/cm3 at 20 °C

storage temp.

2-30°C

SMILES string

[Mg+2].[Cl-].[Cl-]

InChI

1S/2ClH.Mg/h2*1H;/q;;+2/p-2

InChI key

TWRXJAOTZQYOKJ-UHFFFAOYSA-L

Application

Magnesium chloride (MgCl2) can be used as:
  • An additive in the synthesis of aragonite precipitated calcium carbonate (PCC) to control the polymorphism and morphological structure of PCC.[1]
  • An ammonia-selective absorbent to enhance the synthesis of ammonia by Haber−Bosch process.[2]
  • A precursor salt to synthesize magnesium β-ketoesterate complexes by reacting with proligand and anhydrous triethylamine in dry DMF.[3]

Analysis Note

Assay (argentometric): ≥ 98.0 %

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Effects of magnesium chloride and organic additives on the synthesis of aragonite precipitated calcium carbonate
Park WK, et al.
Journal of Crystal Growth, 310(10), 2593-2601 (2008)
Synthesis and structure of anhydrous complexes of magnesium (II) with β-ketoesters of higher alcohols
Koval LI, et al.
Polyhedron, 28(13), 2698-2702 (2009)
Ammonia synthesis enhanced by magnesium chloride absorption
Himstedt HH, et al.
AIChE Journal, 61(4), 1364-1371 (2015)

Questions

1–2 of 2 Questions  
  1. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

  2. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service