Skip to Content
Merck
All Photos(4)

Documents

A9754

Sigma-Aldrich

2-Amino-2-methyl-1,3-propanediol

≥99%

Synonym(s):

AMPD, Ammediol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(HOCH2)2C(NH2)CH3
CAS Number:
Molecular Weight:
105.14
Beilstein:
635708
EC Number:
MDL number:
UNSPSC Code:
12161700
PubChem Substance ID:
NACRES:
NA.25

vapor density

3.63 (vs air)

vapor pressure

~10 mmHg ( 152 °C)

Assay

≥99%

form

crystalline

pH

10.5-11.5 (25 °C, 105.1 g/L)

useful pH range

7.8-9.7

pKa (25 °C)

8.8

bp

151 °C/10 mmHg (lit.)

mp

100-110 °C (lit.)

solubility

H2O: 100 mg/mL, clear to slightly hazy, colorless to very faintly yellow

application(s)

diagnostic assay manufacturing

SMILES string

CC(N)(CO)CO

InChI

1S/C4H11NO2/c1-4(5,2-6)3-7/h6-7H,2-3,5H2,1H3

InChI key

UXFQFBNBSPQBJW-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Buffer component in a SDS-gradient gel electrophoresis system that separates polypeptides in the molecular weight range of 1500 to 100,000. Used as a spacer in isotachophoresis of proteins. Also used as a buffer for the determination of alkaline phosphatase activity.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

>212.0 °F

Flash Point(C)

> 100 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Masahiro Fukuyama et al.
Frontiers in cell and developmental biology, 9, 590333-590333 (2021-08-20)
Current approaches for human hair follicle (HF) regeneration mostly adopt cell-autonomous tissue reassembly in a permissive murine intracorporeal environment. This, together with the limitation in human-derived trichogenic starting materials, potentially hinders the bioengineering of human HF structures, especially for the
Murugappan Sathappa et al.
Bio-protocol, 6(22) (2016-12-09)
Biological membranes are vital for diverse cellular functions such as maintaining cell and organelle structure, selective permeability, active transport, and signaling. The surface charge of the membrane bilayer plays a critical role in these myriad processes. For most biomembranes, the
Karen M O'Connor et al.
Journal of pharmaceutical sciences, 91(10), 2271-2281 (2002-09-13)
Dissolution of diclofenac from compressed discs containing mixtures of a diclofenac salt and a basic excipient, in various w/w ratios, was examined. Two diclofenac salts, diclofenac deanol (DDNL) and diclofenac tert-butylamine, and the basic excipient 2-amino-2-methyl-1,3-propanediol (AMPD) were examined. Inclusion
H Kikuchi et al.
The Tohoku journal of experimental medicine, 173(4), 391-397 (1994-08-01)
The method by MacQueen and Plaut was modified to devise simple microdetermination method of l-lactate which is applicable to deproteinized blood sample. Blood was deproteinized with addition of 25% (w/w) trichloroacetic acid (TCA), and 0.075 M 2-amino-2-methyl-1, 3-propanediole (AMPD) was
Richard E Wooley et al.
Diseases of aquatic organisms, 59(3), 263-267 (2004-07-22)
The antimicrobial agents used to treat bacterial fish diseases are archaic, and their uses may result in the emergence of drug-resistant bacterial strains. This study evaluated the in vitro antimicrobial activity of combinations of Tricide and neomycin or oxytetracycline on

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service