Skip to Content
Merck
All Photos(2)

Documents

225630

Sigma-Aldrich

Tris(2-aminoethyl)amine

96%

Synonym(s):

2,2′,2′′-Nitrilotriethylamine, 2,2′,2′′-Triaminotriethylamine, TAEA

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(NH2CH2CH2)3N
CAS Number:
Molecular Weight:
146.23
Beilstein:
1739626
EC Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

vapor density

5 (vs air)

vapor pressure

0.02 mmHg ( 20 °C)

Assay

96%

form

liquid

refractive index

n20/D 1.497 (lit.)

bp

114 °C/15 mmHg (lit.)

density

0.976 g/mL at 20 °C (lit.)

SMILES string

NCCN(CCN)CCN

InChI

1S/C6H18N4/c7-1-4-10(5-2-8)6-3-9/h1-9H2

InChI key

MBYLVOKEDDQJDY-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Tris(2-aminoethyl)amine (TREN) is a water soluble tripodal ligand that is majorly used in co-ordination chemistry. It has three aminoethylgroups that attach with the surface atoms to provide a scaffold assembly.

Application

Building block for cryptands; precursor to a proazaphosphatrane, a stong, nonionic base.
TREN can be grafted with multi-walled carbon nanotube (MWCNT) for use in solid phase extraction of metal ions for wastewater treatment based applications. It can also be used as a chelating agent in the surface treatment of silica nanoparticles which can further be used for a wide range of industrial applications.

Pictograms

Skull and crossbonesCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 2 Dermal - Acute Tox. 3 Oral - Skin Corr. 1B

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Removal of lead ions from wastewater using functionalized multiwalled carbon nanotubes with tris (2-aminoethyl) amine
Tehrani MS, et al.
Journal of Environmental Protection, 4(06), 529-529 (2013)
Polish Journal of Chemistry, 80, 1825-1825 (2006)
Surface treatment of silica nanoparticles for stable and charge-controlled colloidal silica
Kim K, et al.
International journal of nanomedicine, 9(2), 29-29 (2014)
Living radical polymerization of vinyl chloride initiated with iodoform and catalyzed by nascent Cu0/tris (2-aminoethyl) amine or polyethyleneimine in water at 25? C proceeds by a new competing pathways mechanism.
Percec V, et al.
Journal of Polymer Science Part A: Polymer Chemistry, 41(21), 3283-3299 (2003)
Jung-Ho Shin et al.
Proceedings of the National Academy of Sciences of the United States of America, 108(12), 5045-5050 (2011-03-09)
Zinc is one of the essential transition metals in cells. Excess or lack of zinc is detrimental, and cells exploit highly sensitive zinc-binding regulators to achieve homeostasis. In this article, we present a crystal structure of active Zur from Streptomyces

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service