Skip to Content
Merck
All Photos(1)

Key Documents

29240

Sigma-Aldrich

Cyclohexene

contains 100 ppm BHT as inhibitor, ≥99.0%

Synonym(s):

Tetrahydrobenzene

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H10
CAS Number:
Molecular Weight:
82.14
Beilstein:
906737
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.21
bp:
83 °C (lit.)
vapor pressure:
160 mmHg ( 20 °C)

vapor density

2.8 (vs air)

Quality Level

vapor pressure

160 mmHg ( 20 °C)

Assay

≥99.0%

autoignition temp.

590 °F

contains

100 ppm BHT as inhibitor

expl. lim.

5 %

evapn. residue

≤0.01%

refractive index

n20/D 1.446 (lit.)
n20/D 1.446

bp

83 °C (lit.)

mp

−104 °C (lit.)

density

0.811 g/mL at 25 °C (lit.)

SMILES string

C1CCC=CC1

InChI

1S/C6H10/c1-2-4-6-5-3-1/h1-2H,3-6H2

InChI key

HGCIXCUEYOPUTN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Cyclohexene has been used in the green synthesis of cyclohexene oxide via hydrogen peroxide epoxidation in glycerol-based solvents using bis[3,5-bis(trifluoromethyl)-diphenyl] diselenide as a precatalyst. It can also undergo hydrogen peroxide oxidation in the presence of peroxytungstate-oxalic acid complex catalyst to form adipic acid.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 2 - Asp. Tox. 1 - Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

1.4 °F

Flash Point(C)

-17 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Epoxidation of cyclooctene and cyclohexene with hydrogen peroxide catalyzed by bis [3, 5-bis (trifluoromethyl)-diphenyl] diselenide: Recyclable catalyst-containing phases through the use of glycerol-derived solvents
Garcia-Marin H, et al.
J. Mol. Catal. A: Chem., 334(1), 83-88 (2011)
Partial liquid phase hydrogenation of benzene to cyclohexene over ruthenium catalysts in the presence of an aqueous salt solution: I. Preparation, characterization of the catalyst and study of a number of process variables.
Struijk J, et al.
Applied Catalysis A: General, 83(2), 263-295 (1992)
Hydration of cyclohexene with solid acid catalysts.
Zhang H, et al.
Chemical Engineering Science, 57(2), 315-322 (2002)
Catalytic hydrogenation of cyclohexene: 3. Gas-phase and liquid-phase reaction on supported palladium.
Gonzo EE and Boudart M.
J. Catal., 52(3), 462-471 (1978)
Hikaru Yanai et al.
Chemical communications (Cambridge, England), 47(25), 7245-7247 (2011-05-25)
In the absence of any additional catalysts, the reactions of (CF(3)SO)(2)CH(2), aldehydes, and 1,3-dienes gave gem-bis(triflyl)cyclohexenes in excellent yields with high regioselectivity. gem-Bis(triflyl)cyclohexene products can be easily converted to the corresponding aryl trifluoromethyl sulfones.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service