Skip to Content
Merck
All Photos(2)

Key Documents

8.22328

Sigma-Aldrich

Cyclohexanol

for synthesis

Synonym(s):

Hexahydrophenol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H11OH
CAS Number:
Molecular Weight:
100.16
Beilstein:
906744
MDL number:
UNSPSC Code:
12352104
EC Index Number:
203-630-6
NACRES:
NA.22
grade:
for synthesis
synthesis grade
bp:
160-161 °C (lit.)
vapor pressure:
0.98 mmHg ( 25 °C)

grade

for synthesis
synthesis grade

Quality Level

vapor density

3.5 (vs air)

vapor pressure

0.98 mmHg ( 25 °C)

Assay

≥99% (GC)

form

solid

autoignition temp.

572 °F

potency

1400 mg/kg LD50, oral (Rat)

expl. lim.

12.25 %

dilution

(for synthesis)

refractive index

n20/D 1.465 (lit.)

pH

6.5 (20 °C, 40 g/L in H2O)

bp

160-161 °C (lit.)

mp

20-22 °C (lit.)

solubility

40 g/L

density

0.948 g/mL at 25 °C (lit.)

storage temp.

2-30°C

SMILES string

OC1CCCCC1

InChI

1S/C6H12O/c7-6-4-2-1-3-5-6/h6-7H,1-5H2

InChI key

HPXRVTGHNJAIIH-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Analysis Note

Assay (GC, area%): ≥ 99.0 % (a/a)
Water (K. F.): ≤ 0.20 %
Identity (IR): passes test
Due to its specific melting range the product may be solid, liquid, a solidified melt or a supercooled melt.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 3 - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

147.2 °F - closed cup - DIN 51794

Flash Point(C)

64 °C - closed cup - DIN 51794


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service