Skip to Content
Merck
All Photos(2)

Key Documents

125415

Sigma-Aldrich

1,4-Cyclohexadiene

97%

Synonym(s):

1,4-Dihydrobenzene

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H8
CAS Number:
Molecular Weight:
80.13
Beilstein:
1900733
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

97%

form

liquid

contains

~0.1% hydroquinone as stabilizer

impurities

3% benzene

refractive index

n20/D 1.472 (lit.)

bp

88-89 °C (lit.)

density

0.847 g/mL at 25 °C (lit.)

storage temp.

2-8°C

SMILES string

C1C=CCC=C1

InChI

1S/C6H8/c1-2-4-6-5-3-1/h1-2,5-6H,3-4H2

InChI key

UVJHQYIOXKWHFD-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

1,4-Cyclohexadiene is an effective hydrogen donor for catalytic hydrogenation reactions. It can rapidly replace benzyl groups of N-benzyloxycarbamates, benzyl esters, benzyl ethers and benzyl amines with hydrogen. It forms benzene at elevated temperatures in the presence of a ruthenium(II)-triphenylphosphine catalyst.

Application

1,4-Cyclohexadiene (1,4-CHD) was used to study the formation of parent ion from heavy fragmentation of 1,4-CHD on irradiation with a high-intensity laser pulse.
Useful for the reduction of radical intermediates formed in electron-transfer mediated ring-opening reactions

Pictograms

FlameHealth hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Carc. 1A - Flam. Liq. 2 - Muta. 1B - STOT RE 2

Target Organs

Blood

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

19.4 °F - closed cup

Flash Point(C)

-7 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A key factor in parent and fragment ion formation on irradiation with an intense femtosecond laser pulse.
Harada H, et al.
Chemical Physics Letters, 342(5), 563-570 (2001)
Organometallics, 25, 5456-5456 (2006)
Kazutada Ikeuchi et al.
Organic letters, 14(23), 6016-6019 (2012-11-15)
Asymmetric bromolactonization of prochiral cyclohexadiene derivatives with N-bromosuccimide proceeded in the presence of (DHQD)(2)PHAL as a chiral catalyst to afford the corresponding bromolactones with up to 93% ee. This reaction was also applicable to the kinetic resolution of a racemic
Kyung-Bin Cho et al.
Chemistry (Weinheim an der Bergstrasse, Germany), 18(33), 10444-10453 (2012-06-21)
The experimentally measured bimolecular reaction rate constant, k(2), should in principle correlate with the theoretically calculated rate-limiting free energy barrier, ΔG(≠), through the Eyring equation, but it fails quite often to do so due to the inability of current computational
Jaya S Kudavalli et al.
Organic letters, 12(23), 5550-5553 (2010-11-09)
Benzene-cis- and trans-1,2-dihydrodiols undergo acid-catalyzed dehydration at remarkably different rates: k(cis)/k(trans) = 4500. This is explained by formation of a β-hydroxycarbocation intermediate in different initial conformations, one of which is stabilized by hyperconjugation amplified by an aromatic no-bond resonance structure

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service