Skip to Content
Merck
All Photos(3)

Documents

222550

Sigma-Aldrich

Nile Blue chloride

Dye content 85 %

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H20ClN3O
CAS Number:
Molecular Weight:
353.85
Colour Index Number:
51180
Beilstein:
3923952
MDL number:
UNSPSC Code:
12171500
PubChem Substance ID:
NACRES:
NA.47

form

powder

Quality Level

composition

Dye content, 85%

solubility

water: 1 mg/mL, opaque, blue to very deep blue

λmax

638 nm

application(s)

diagnostic assay manufacturing
hematology
histology

storage temp.

room temp

SMILES string

[Cl-].CC\[N+](CC)=C1/C=CC2=Nc3c(OC2=C1)cc(N)c4ccccc34

InChI

1S/C20H19N3O.ClH/c1-3-23(4-2)13-9-10-17-18(11-13)24-19-12-16(21)14-7-5-6-8-15(14)20(19)22-17;/h5-12,21H,3-4H2,1-2H3;1H

InChI key

SHXOKQKTZJXHHR-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Nile blue chloride has been used to stain non-aqueous phase in fluorescence microscopy studies.

Biochem/physiol Actions

Nile blue chloride is a green colored powder and is a member of oxazine group of dyes. It changes its color from blue (pH 9.4) to purple-red (11.0). Nile blue chloride has been used to detect DNA and lipids. In addition, it also helps to identify bacterial growth. Nile blue chloride also facilitates diagnosis of disease linked with amyloid accumulation.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Effect of ?-casein on surface activity of Quillaja bark saponin at fluid/fluid interfaces
Wojciechowski K
Food Hydrocolloids, 34, 208-216 (2014)
Jason D Slinker et al.
Nature chemistry, 3(3), 228-233 (2011-02-22)
Molecular wires show promise in nanoscale electronics, but the synthesis of uniform, long conductive molecules is a significant challenge. Deoxyribonucleic acid (DNA) of precise length, by contrast, is synthesized easily, but its conductivity over the distances required for nanoscale devices
R. W. Sabnis
Handbook of Acid-Base Indicators (2007)
Julie Dutet et al.
Pharmaceutical research, 29(12), 3464-3474 (2012-08-08)
To estimate the in vitro ungual penetration depth of sodium fluorescein and nile blue chloride by laser scanning confocal microscopy. The depth, uniformity and pathways of penetration of both markers into human nail during passive and iontophoretic experiments were investigated.
Zhiping Cui et al.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy, 83(1), 1-7 (2011-09-29)
A dual-wavelength overlapping resonance Rayleigh scattering (DWO-RRS) method was developed to detect chondroitin sulfate (CS) with nile blue sulfate (NBS). At pH 3.0-4.0 Britton-Robinson (BR) buffer medium, CS interacted with NBS to form an ion-association complex. As a result, the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service