Skip to Content
Merck
All Photos(1)

Documents

370088

Sigma-Aldrich

Nile Blue A perchlorate

Dye content 95 %

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H20ClN3O5
CAS Number:
Molecular Weight:
417.84
EC Number:
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

form

powder

composition

Dye content, 95%

mp

290-292 °C (dec.) (lit.)

λmax

628 nm

SMILES string

[O-]Cl(=O)(=O)=O.CC\[N+](CC)=C1/C=CC2=Nc3c(OC2=C1)cc(N)c4ccccc34

InChI

1S/C20H19N3O.ClHO4/c1-3-23(4-2)13-9-10-17-18(11-13)24-19-12-16(21)14-7-5-6-8-15(14)20(19)22-17;2-1(3,4)5/h5-12,21H,3-4H2,1-2H3;(H,2,3,4,5)

InChI key

OUAWJLNOTQAOHL-UHFFFAOYSA-N

General description

Nile Blue A perchlorate (NBA) is a cationic oxazine dye that has properties of both the electron donor and acceptor. It contains two amine groups that form a push-pull conjugating module. Its sensitivity to the environment allows it to have the solvatochromic characteristic.

Application

NBA can be used in the confocal microscopy of imidazolium functionalized clay. It can also be used in the labeling of silver nanoparticles for surface enhanced raman scattering (SERS). Silica gel doped with NBA can be potentially used as a solid state dye for laser based materials.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Characterizing the spatial dependence of redox chemistry on plasmonic nanoparticle electrodes using correlated super-resolution surface-enhanced Raman scattering imaging and electron microscopy.
Weber ML, et al.
The Journal of Physical Chemistry C, 119(32), 18591-18601 (2015)
Synthesis and spectroscopic characterization of Nile Blue doped silica gel rods.
Haque FZ, et al.
Optik, 124(20), 4287-4291 (2013)
The photophysical properties of Nile red and Nile blue in ordered anisotropic media.
Tajalli H, et al.
Dyes and Pigments, 78(1), 15-24 (2008)
Synthesis of imidazolium salts and their application in epoxy montmorillonite nanocomposites.
Langat J, et al.
Polymer, 47(19), 6698-6709 (2006)
Julie Dutet et al.
Pharmaceutical research, 29(12), 3464-3474 (2012-08-08)
To estimate the in vitro ungual penetration depth of sodium fluorescein and nile blue chloride by laser scanning confocal microscopy. The depth, uniformity and pathways of penetration of both markers into human nail during passive and iontophoretic experiments were investigated.

Articles

Fluorescence quenching microscopy visualizes 2D materials like graphene and MoS2 rapidly, inexpensively, and with high fidelity.

Fluorescence quenching microscopy visualizes 2D materials like graphene and MoS2 rapidly, inexpensively, and with high fidelity.

Fluorescence quenching microscopy visualizes 2D materials like graphene and MoS2 rapidly, inexpensively, and with high fidelity.

Fluorescence quenching microscopy visualizes 2D materials like graphene and MoS2 rapidly, inexpensively, and with high fidelity.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service