Skip to Content
Merck
Nile Blue chloride Dye content 85 %

Nile Blue chloride

Nile Blue chloride Dye content 85 %
Empirical Formula (Hill Notation):
C20H20ClN3O
CAS Number:
Molecular Weight:
353.85

Products

Product Category
Brand
Formula Weight
Physical Form
Chemical Composition
Quality Segment
Application
Showing 1-1 of 1
Compare
Product Number
Description
Pricing