Skip to Content
Merck
All Photos(2)

Key Documents

19533

Supelco

Geranyl pyrophosphate lithium salt

analytical standard

Synonym(s):

(E)-3,7-Dimethyl-2,6-octadien-1-ol trihydrogen pyrophosphate lithium salt, (E)-3,7-Dimethyl-2,6-octadien-1-yl pyrophosphate lithium salt, GPP-Li

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H20O7P2 · xLi+
CAS Number:
Molecular Weight:
314.21 (free acid basis)
Beilstein:
1915690
MDL number:
UNSPSC Code:
85151701
NACRES:
NA.24

Quality Level

grade

analytical standard

Assay

≥95.0% (HPLC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

food and beverages

format

neat

shipped in

dry ice

storage temp.

−20°C

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Analysis Note

NMR-Internal Standard : KHP

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Akira Saito et al.
Melanoma research, 18(2), 85-94 (2008-03-14)
Competitive inhibitors of 3-hydroxy-3-methylglutaryl coenzyme A reductase (the statins) that inhibit the synthesis of mevalonic acid are in wide use for treatment of hypercholesterolemia. Although antitumor effects on a variety of cell types have been reported for statins, the effect
Fu-Lien Hsieh et al.
Journal of molecular biology, 404(5), 859-873 (2010-10-23)
Isoprenoids, most of them synthesized by prenyltransferases (PTSs), are a class of important biologically active compounds with diverse functions. The mint geranyl pyrophosphate synthase (GPPS) is a heterotetramer composed of two LSU·SSU (large/small subunit) dimers. In addition to C(10)-GPP, the
Ravi S Singh et al.
BMC molecular biology, 11, 88-88 (2010-11-26)
Geranyl pyrophosphate (GPP) and p-hydroxybenzoate (PHB) are the basic precursors involved in shikonins biosynthesis. GPP is derived from mevalonate (MVA) and/or 2-C-methyl-D-erythritol 4-phosphate (MEP) pathway(s), depending upon the metabolite and the plant system under consideration. PHB, however, is synthesized by
K C Chang et al.
The Journal of experimental zoology, 290(6), 642-651 (2001-12-19)
BALB/3T3 cells were transformed by transfection with DNA encoding the mutated ras(Q(61)K) from shrimp Penaeus japonicus (Huang et al., 2000). The GTPase-activating protein (GAP) in the cytosol fraction was significantly expressed and degraded, compared to untransformed cells on the western
Sarah A Holstein et al.
Lipids, 39(4), 293-309 (2004-09-11)
The isoprenoid biosynthetic pathway is the source of a wide array of products. The pathway has been highly conserved throughout evolution, and isoprenoids are some of the most ancient biomolecules ever identified, playing key roles in many life forms. In

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service