Skip to Content
Merck
All Photos(1)

Key Documents

442785

Supelco

Squalene

analytical standard

Synonym(s):

2,6,10,15,19,23-Hexamethyl-2,6,10,14,18,22-tetracosahexaene

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[(CH3)2C[=CHCH2CH2C(CH3)]2=CHCH2-]2
CAS Number:
Molecular Weight:
410.72
Beilstein:
1728919
EC Number:
MDL number:
UNSPSC Code:
12000000
PubChem Substance ID:

grade

analytical standard

CofA

current certificate can be downloaded

packaging

ampule of 1000 mg

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

refractive index

n20/D 1.494 (lit.)

bp

285 °C/25 mmHg (lit.)

mp

−75 °C (lit.)

density

0.858 g/mL at 25 °C (lit.)

application(s)

environmental
food and beverages

format

neat

storage temp.

2-30°C

SMILES string

CC(C)=CCCC(C)=CCCC(C)=CCC\C=C(/C)CCC=C(C)CCC=C(C)C

InChI

1S/C30H50/c1-25(2)15-11-19-29(7)23-13-21-27(5)17-9-10-18-28(6)22-14-24-30(8)20-12-16-26(3)4/h15-18,23-24H,9-14,19-22H2,1-8H3/b27-17+,28-18+,29-23+,30-24+

InChI key

YYGNTYWPHWGJRM-AAJYLUCBSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Squalene is a naturally occurring triterpenoid and a precursor to sterol biosynthesis. Typically isolated from shark liver, olive, amaranth, and algal oils, it shows various cosmetic and medicinal uses.

Application

This analytical standard can also be used as follows:

  • Analysis of 119 vegetable oil samples from 7 different varieties to detect and determine squalene and four sterols using a developed and validated non-destructive method based on proton nuclear magnetic resonance (1H NMR) spectroscopy combined with partial least squares (PLS) method
  • Determination of squalene in different oil samples by gas chromatography coupled to flame ionization detection (GC-FID) following base-catalyzed transmethylation of the oil base
  • Ultra-high performance liquid chromatography-photodiode array detection (UHPLC-PDA) based quantification of squalene, following its single-step solid phase extraction (SPE) from 33 samples of extra virgin olive oil collected from different countries
  • Measurement of squalene content of 16 commercial shark liver oil samples using a gas chromatography-mass spectrometry-based method, in addition to IR and Raman spectroscopy combined with the PLS method
  • Multi-residue determination of 14 naturally occurring compounds— squalene, tocopherols, and phytosterols, following their solid phase extraction (SPE) from 15 vegetable oil samples by GC-MS

Other Notes

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Asp. Tox. 1

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

F Rubinstein et al.
BMJ (Clinical research ed.), 346, f393-f393 (2013-02-06)
To assess the risk of adverse perinatal events of vaccination of pregnant women with an MF59 adjuvanted vaccine. Cross sectional multicentre study. 49 public hospitals in major cities in Argentina, from September 2010 to May 2011. 30,448 mothers (7293 vaccinated)
L Harivardhan Reddy et al.
Advanced drug delivery reviews, 61(15), 1412-1426 (2009-10-07)
Squalene is a natural lipid belonging to the terpenoid family and a precursor of cholesterol biosynthesis. It is synthesized in humans and also in a wide array of organisms and substances, from sharks to olives and even bran, among others.
Alicja M Chmielewska et al.
Journal of virology, 88(10), 5502-5510 (2014-03-07)
Despite the recent progress in the development of new antiviral agents, hepatitis C virus (HCV) infection remains a major global health problem, and there is a need for a preventive vaccine. We previously reported that adenoviral vectors expressing HCV nonstructural
Christine Durier et al.
AIDS (London, England), 27(1), 87-93 (2012-09-29)
In immunocompromised patients, alternative schedules more immunogenic than the standard influenza vaccine regimen are necessary to enhance and prolong vaccine efficacy. We previously reported that the AS03A-adjuvanted 2009 A/H1N1v vaccine yielded a higher short-term immune response than the nonadjuvanted one
Joachim Caron et al.
Advanced healthcare materials, 2(1), 172-185 (2012-12-06)
A series of new lipid prodrugs of paclitaxel, which can be formulated as nanoassemblies, are described. These prodrugs which are designed to overcome the limitations due to the systemic toxicity and low water solubility of paclitaxel consist of a squalene

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service