Skip to Content
Merck
All Photos(2)

Key Documents

718467

Sigma-Aldrich

Titanium(IV) oxide

nanopowder, 21 nm primary particle size (TEM), ≥99.5% trace metals basis

Synonym(s):

TiO2 nanopowder, Titania, Titanium dioxide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
TiO2
CAS Number:
Molecular Weight:
79.87
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23

Quality Level

Assay

≥99.5% trace metals basis

form

nanopowder

surface area

35-65 m2/g , BET

primary particle size

21 nm (TEM)

mp

1850 °C
>350 °C (lit.)

density

4.26 g/mL at 25 °C (lit.)

SMILES string

O=[Ti]=O

InChI

1S/2O.Ti

InChI key

GWEVSGVZZGPLCZ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Our titanium(IV) oxide nanopowder is a fine white powder composed of titanium dioxide nanoparticles with an average particle size of 21 nm, resulting in a high surface area of 35-65 m2/g. Two main physico-chemically distinct polymorphs of TiO2 are anatase and rutile. Anatase has a higher photocatalytic activity than rutile but is thermodynamically less stable.

Application

TiO2 nanoparticles were used to study the photocatalytic hexane vapor degradation. It has been used as a sorbent for arsenic removal. TiO2 nanoparticles are suitable for remediation of antiseptic components in wastewater by photocatalysis. It has also been used to study adsorption of DNA oligonucleotides by titanium dioxide nanoparticles.

Storage Class Code

13 - Non Combustible Solids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Xu Zhang et al.
Langmuir : the ACS journal of surfaces and colloids, 30(3), 839-845 (2014-01-07)
Titanium dioxide (TiO2) or titania shows great promise in detoxification and drug delivery. To reach its full potential, it is important to interface TiO2 with biomolecules to harness their molecular recognition function. To this end, DNA attachment is an important
Photocatalytic degradation of hexane vapors in batch and continuous systems using impregnated ZnO nanoparticles
Saucedo-Lucero JO and Arriaga S
Chemical Engineering Journal, 218, 358-367 (2013)
Removal of arsenic from aqueous solution by two types of nano TiO2 crystals.
Ma L and Tu SX
Environmental Chemistry Letters, 9(4), 465-472 (2011)
Remediation of Antiseptic Components in Wastewater by Photocatalysis Using TiO2 Nanoparticles
Das R, et al.
Industrial & Engineering Chemistry Research, 53(8), 3012-3020 (2014)
Shanshan Zhang et al.
Archives of toxicology, 94(4), 1173-1190 (2020-03-13)
Titanium dioxide nanoparticles (TiO2NPs) have been widely used as food additives in daily life. However, the impact of oral intake of TiO2NPs on the nervous system is largely unknown. In this study, 7-week-old mice were treated with either vehicle or

Articles

Dye-sensitized solar cells directly convert sunlight to electricity

Dye-sensitized solar cells (DSSCs) attract attention for high performance and potential low-cost production in solar energy.

Dye-sensitized solar cells (DSSCs) attract attention for high performance and potential low-cost production in solar energy.

Dye-sensitized solar cells (DSSCs) attract attention for high performance and potential low-cost production in solar energy.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service