Skip to Content
Merck
All Photos(2)

Key Documents

17929

Sigma-Aldrich

O-Methyl-O′-succinylpolyethylene glycol 5′000

Synonym(s):

Polyethylene glycol, O-(Succinyl)-O′-methylpolyethylene glycol 5′000, Polyethylene glycol 5000 monomethyl ether succinate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3O(CH2CH2O)nCOCH2CH2COOH
CAS Number:
MDL number:
UNSPSC Code:
51171641
NACRES:
NA.26

form

powder

Quality Level

mol wt

Mr ~5100

mp

56-60 °C

Ω-end

carboxylic acid

α-end

methoxy

storage temp.

−20°C

Application

Polyethylene glycol 5000 is used in the construction of biomaterials such as biopolymers, micelles and nanostructures.
Soluble polymer for solid phase synthesis of imines and β-lactams; attaching peptides to PEG

Biochem/physiol Actions

O-Methyl-O′-succinylpolyethylene glycol 5′000 is a modified monomethoxypolyethylene glycol (mPEG) wherein the hydroxyl groups are succinylated by succinic anhydride. This succinylated mPEG can then be used to synthesize tolerogenic mPEG conjugates of peptides. Tolerogenic peptide conjugates are very effective reagents for obtaining epitope-specific immunosuppression of antibody responses to immunopathogenic sites on multideterminant complex protein antigens.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

M. Joppich, P.L. Luisi
Makromol. Chem., 180, 1381-1381 (1979)
V. Molteni et al.
Tetrahedron Letters, 39, 1257-1257 (1998)
A Bogdanov et al.
Journal of drug targeting, 4(5), 321-330 (1997-01-01)
A co-polymer of O-methyl polyethylene(glycol)-O'-succinate (MPEGs, m.w. 5100) and poly-l-lysine (PL, median m.w. 32700, degree of polymerization 256) has been synthesized by covalent grafting. The resultant MPEGs-PL (30% modification degree of epsilon-amino groups) had a hydrodynamic diameter corresponding to a
Mario Beyer et al.
Methods in molecular biology (Clifton, N.J.), 570, 309-316 (2009-08-04)
Combinatorial synthesis of peptides on solid supports (1), either as spots on cellulose membranes (2) or with split-pool-libraries on polymer beads (3), substantially forwarded research in the field of peptide-protein interactions. Admittedly, these concepts have specific limitations, on one hand
Telli Hekmatara et al.
Journal of nanoscience and nanotechnology, 9(8), 5091-5098 (2009-11-26)
Hydrophobic drugs, loperamide and paclitaxel, were loaded in poly(butyl cyanoacrylate) nanoparticles by polymerization of n-butyl-2-cyanoacrylate in aqueous-organic media in the presence of a drug. The particles were stabilized by dextran 70,000 and poloxamer 188 or by 1,2-dioleoyl-sn-glycero-3-phosphoethanolamine-N-[methoxy(polyethylene glycol)-5000] sodium salt.

Articles

Kanjiro Miyata (The University of Tokyo, Japan) provides insights on the rational design of polymeric materials for “smart” oligonucleotide delivery.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service