Skip to Content
Merck
All Photos(1)

Key Documents

528877

Millipore

Polyethylene glycol

6000, Molecular Biology Grade

Synonym(s):

Poly(ethylene glycol), Polyethylene Glycol 6000, PEG

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
H(OCH2CH2)nOH
CAS Number:
MDL number:
UNSPSC Code:
51171641
NACRES:
NA.25
Pricing and availability is not currently available.

grade

Molecular Biology

Quality Level

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

color

white

solubility

water: 200 mg/mL

cation traces

heavy metals (as Pb): ≤0.0005%

absorption

≤0.1 at 280 nm
≤0.2 at 260 nm

foreign activity

DNase, proteases, RNase, none detected

shipped in

ambient

storage temp.

15-25°C

SMILES string

C(CO)O

InChI

1S/C2H6O2/c3-1-2-4/h3-4H,1-2H2

InChI key

LYCAIKOWRPUZTN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Useful as a stationary phase in gas chromatography (absorbance: ≤0.2 at 260 nm; ≤0.1 at 280 nm).

Application


  • Challenges in Pharmacokinetic Modelling of [(18)F]fluoro-PEG-folate PET/CT Imaging in Epithelial Ovarian Cancer Patients.: This study addresses the complexities of pharmacokinetic modeling in using Polyethylene glycol (PEG)-conjugated imaging agents for PET/CT in ovarian cancer, highlighting the adaptability and clinical implications of PEG in diagnostic imaging (Ruytenberg et al., 2024).

  • Assessing the toxicity of one-step-synthesized PEG-coated gold nanoparticles: in vitro and in vivo studies.: Research into the biocompatibility of PEG-coated gold nanoparticles synthesized in a single step, evaluating their safety profile across both cellular and animal models, thus underscoring PEG′s role in enhancing nanomedicine safety (Garrigós et al., 2024).

  • Microwave synthesized novel biomass carbon dots applied in the fluorescent detection of crystal violet.: The study explores the use of PEG in the synthesis of carbon dots for enhanced fluorescence in dye detection, offering insights into the broader applicative potential of PEG in environmental monitoring and safety (Yin et al., 2024).

  • Modified zero-valent iron nanoparticles enhanced remediation of PCBs-contaminated soil.: Illustrating PEG′s utility in environmental cleanup, this study details the enhancement of soil remediation techniques through the application of PEG-modified nanoparticles, reflecting PEG′s versatility across different sectors (Zhao et al., 2024).

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, store at room temperature (20°C). Stock solutions are stable for up to 6 months at room temperature.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

281.5 °F - closed cup

Flash Point(C)

138.6 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service