Skip to Content
Merck
All Photos(3)

Key Documents

81323

Sigma-Aldrich

Poly(ethylene glycol) methyl ether

average MN 5,000, methoxy, hydroxyl

Synonym(s):

Polyethylene glycol, Methoxy poly(ethylene glycol), Polyethylene glycol monomethyl ether, mPEG

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(OCH2CH2)nOH
CAS Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

Product Name

Poly(ethylene glycol) methyl ether, average Mn 5,000

vapor density

>1 (vs air)

vapor pressure

0.05 mmHg ( 20 °C)

form

flakes
powder or crystals

mol wt

average Mn 5,000

mp

60-64 °C

Ω-end

hydroxyl

α-end

methoxy

SMILES string

O(CCO)C

InChI

1S/C3H8O2/c1-5-3-2-4/h4H,2-3H2,1H3

InChI key

XNWFRZJHXBZDAG-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Poly(ethylene glycol) methyl ether (mPEG) is a hydrophilic polymer that is used to control the flexibility of a composite. mPEG can be used for a variety of applications such as drug delivery, tissue engineering, and other biological uses.

Other Notes

Polymer used in the polymer-supported liquid synthesis of oligosaccharides

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

359.6 °F - closed cup

Flash Point(C)

182 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Magnetite nanoparticles stabilized with polymeric bilayer of poly (ethylene glycol) methyl ether-poly (?-caprolactone) copolymers
Meerod S, et al.
Polymer, 49(18), 3950-3956 (2008)
S.P. Douglas et al.
Journal of the American Chemical Society, 113, 5095-5095 (1991)
Hamed Nosrati et al.
Pharmaceutical development and technology, 24(1), 89-98 (2018-01-09)
In this study, we designed a polymersome system for the controlled release of methotrexate (MTX) as an anticancer drug with the objective of improving the loading efficiency of the drug in polymersomes as well as achievement of an efficient control
Kangan Li et al.
International journal of nanomedicine, 8, 2589-2600 (2013-07-28)
Development of dual-mode or multi-mode imaging contrast agents is important for accurate and self-confirmatory diagnosis of cancer. We report a new multifunctional, dendrimer-based gold nanoparticle (AuNP) as a dual-modality contrast agent for magnetic resonance (MR)/computed tomography (CT) imaging of breast
Yvonne K Girard et al.
PloS one, 8(10), e75345-e75345 (2013-10-23)
The development of a suitable three dimensional (3D) culture system for anticancer drug development remains an unmet need. Despite progress, a simple, rapid, scalable and inexpensive 3D-tumor model that recapitulates in vivo tumorigenesis is lacking. Herein, we report on the

Articles

Biofouling control essential for device performance and safety; minimize accumulation of biomolecules and bioorganisms.

Biofouling control essential for device performance and safety; minimize accumulation of biomolecules and bioorganisms.

Biofouling control essential for device performance and safety; minimize accumulation of biomolecules and bioorganisms.

Biofouling control essential for device performance and safety; minimize accumulation of biomolecules and bioorganisms.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service