Skip to Content
Merck
All Photos(1)

Key Documents

08650

Supelco

Chlorobenzene

analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H5Cl
CAS Number:
Molecular Weight:
112.56
Beilstein:
605632
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

vapor density

3.86 (vs air)

vapor pressure

11.8 mmHg ( 25 °C)

Assay

≥99.7% (GC)

autoignition temp.

1178 °F

shelf life

limited shelf life, expiry date on the label

expl. lim.

7.1 %

refractive index

n20/D 1.524 (lit.)
n20/D 1.525

bp

132 °C (lit.)

mp

−45 °C (lit.)

density

1.106 g/mL at 25 °C (lit.)

application(s)

cleaning products
cosmetics
environmental
food and beverages
personal care

format

neat

SMILES string

Clc1ccccc1

InChI

1S/C6H5Cl/c7-6-4-2-1-3-5-6/h1-5H

InChI key

MVPPADPHJFYWMZ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Pure chlorobenzene is an aromatic organic compound most commonly used as a solvent and as a precursor in chemical syntheses. It is an intermediate in dyes and pesticide production and used in insect repellents, deodorants, and degreasers.

Application

Chlorobenzene may be used as an extraction solvent in the determination of chlorophenols in water samples and triazine herbicides in aqueous samples by chromatography techniques.

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Inhalation - Aquatic Chronic 2 - Flam. Liq. 3 - Skin Irrit. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

80.6 °F

Flash Point(C)

27 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 2

1 of 2

Determination of triazine herbicides in aqueous samples by dispersive liquid-liquid microextraction with gas chromatography-ion trap mass spectrometry
Nagaraju D and Huang SD
Journal of Chromatography A, 1161(1-2), 89-97 (2007)
In situ crystallization and crystal structure determination of chlorobenzene
Nath NK and Naumov P
Macedonian Journal of Chemistry and Chemical Engineering, 34(1), 63-66 (2015)
Determination of chlorobenzenes in water by drop-based liquid-phase microextraction and gas chromatography-electron capture detection
Tor A
Journal of Chromatography A, 1125(1), 129-132 (2006)
Determination of chlorophenols in water samples using simultaneous dispersive liquid--liquid microextraction and derivatization followed by gas chromatography-electron-capture detection.
Fattahi N, et al.
Journal of Chromatography A, 1157(1-2), 23-29 (2007)
Kyeongil Hwang et al.
Advanced materials (Deerfield Beach, Fla.), 27(7), 1241-1247 (2015-01-13)
Fully printed perovskite solar cells are demonstrated with slot-die coating, a scalable printing method. A sequential slot-die coating process is developed to produce efficient perovskite solar cells and to be used in a large-scale roll-to-roll printing process. All layers excluding

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service